Applied Biosystems™

Clariom™ S Pico Assay HT, human

Catalog number: 902963
Have Questions?
Applied Biosystems™

Clariom™ S Pico Assay HT, human

Catalog number: 902963
Have Questions?
Catalog Number
902963
Unit Size
30 reactions
Price (USD)
Catalog Number
Unit Size
96 reactions
Price (USD)
Catalog NumberUnit SizePrice (USD)
90296330 reactions
90296496 reactions
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Obtain a gene-level view of the human transcriptome with Clariom S Pico Assays HT for human samples. Clariom S Pico Assays serve as a next generation transcriptome-wide gene-level expression profiling tool, which allow for the fastest, simplest, and most scalable path to generating the results you need for your research. Based on industry-leading microarray technology, the novel Human HT Clariom S Assay design provides extensive coverage of all known well-annotated genes, compatibility with research sample types, scalable formats, and flexible data analysis software. Clariom S Pico Assays HT are the tools of choice to find expression biomarkers with known function as quickly, easily, and cost-effectively as possible.

Find answers, move on
Although the number of known transcribed genes has expanded rapidly in recent years, knowledge of the function of each gene is still evolving. Many genes and transcripts found in databases are poorly annotated or unannotated, which can complicate and prolong data analysis and interpretation. Human HT Clariom S Pico Assays focus on well-annotated genes, providing researchers with the ability to perform gene-level expression profiling studies and to quickly assess changes in key genes and pathways. With less time required for data analysis, Clariom S Pico Assays HT for human help researchers reach conclusions more rapidly.

Simple, swift biomarker discovery
• Accurately measure gene-level expression from >20,000 well-annotated genes to get to answers quickly.
• Choose a format that suits your throughput needs, processing from 1 to 192 samples a day.
• Go from data to insight in minutes with intuitive, highly visual, free analysis software designed for the biologist.

When you have precious samples, get it right the first time
• Generate robust expression profiles from as little as 100 pg of total RNA–as few as 10 cells.
• Utilize RNA from various sample types including blood, cells, and fresh/fresh-frozen or FFPE tissues.
• Preserve sample integrity and reduce data variability with an assay that does not require a globin or rRNA removal step.
• Save time and money with fully automated sample preparation options.
Clariom S solutions are available formats for single-sample (cartridge array) processing on the GeneChip™ 3000 instrument system and high-throughput automated processing (plate array) on the GeneTitan™ HT Microarray System, offering the flexibility to accommodate both small and large cohort studies. The complete solution comes with reagents and fast, simple Transcriptome Analysis Console (TAC) software to analyze and visualize global expression patterns of genes, pathways, and network interactions in minutes.

Get the truest level of gene-level expression
To generate robust gene-level expression, Human HT Clariom S Assays detect only the exons present in all known transcript isoforms expressed from a single gene locus-constitutive exons. This differs from other gene-level array technologies and shallow RNA-Seq, which provide either a biased view of gene expression or data that are complicated by variation in expression of transcript variants. By detecting only constitutive exons throughout the length of each known gene, Human HT Clariom S Assays generate the most accurate and truest measurement of gene-level expression available today.

Keeping biomarker identification across the transcriptome simple and swift, Clariom S Pico Assays HT for human provide you with the coverage you require, the reproducibility you need, and the insights you want to act on your discoveries.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Array Format
24-array Plate
Array
Transcriptome Profiling
Sample Type
RNA
For Use With (Application)
Microarray Analysis
No. of Reactions
30
Product Line
Applied Biosystems™
Number of Arrays
24 arrays
Product Type
S Pico Assay HT
Shipping Condition
Approved for shipment at Room Temperature or on Wet or Dry Ice
Species
Human

Contents & Storage

• Control HeLa RNA, store at -20°C
• Hybridization Control Kit, store at -20°C
• IVT Pico Amplification Kit, Module 1, store at -20°C
• IVT Pico Amplification Kit, Module 2, store at -20°C
• IVT Pico Module 3, store at 4°C
• Clariom S 24-Array Plate , human, store at 4°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text