Search Thermo Fisher Scientific
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Catalog Number | Volume (Metric) | Filtered | For Use With (Equipment) | Fit for Purpose |
---|---|---|---|---|
904-262 | 1000 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
901-021 | 10 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
901-251 | 20 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
901-252 | 20 μL | Non-filtered | Cavro, Tecan Genesis, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
901-253 | 20 μL | Non-filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
901-261 | 20 μL | Non-filtered | Cavro, Tecan Genesis, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
901-262 | 20 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
902-011 | 50 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
902-021 | 50 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
902-251 | 50 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
902-252 | 50 μL | Non-filtered | Cavro, Tecan Genesis, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
902-253 | 50 μL | Non-filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
902-261 | 50 μL | Non-filtered | Cavro, Tecan Genesis, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
902-262 | 50 μL | Non-filtered | Tecan Aquarius, Tecan TeMo, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
903-011 | 175 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
903-011G | 175 μL | Filtered | Cavro, MiniPrep | Sterile and PCR Ready |
903-021 | 175 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
903-033 | 175 μL | Filtered | Tecan Freedom EVO LiHa Head, Tecan Genesis LiHa Head | PCR Ready |
903-251 | 200 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
903-251G | 200 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
903-252 | 200 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
903-253 | 200 μL | Non-filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
903-261 | 200 μL | Non-filtered | Tecan Aquarius, Tecan TeMo, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
903-262 | 200 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
904-011 | 1000 μL | Filtered | Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
904-011G | 1000 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
904-013 | 1000 μL | Filtered | Tecan Freedom EVO LiHa Head, Tecan Genesis | PCR Ready |
904-021 | 1000 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
904-251 | 1000 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
904-251G | 1000 μL | Non-filtered | Cavro, Tecan Genesis, Tecan Freedom EVO, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
904-252 | 1000 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
904-252G | 1000 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
904-253 | 1000 μL | Non-filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
904-261 | 1000 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
901-011 | 10 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
903-252G | 200 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
Share catalog number, name or link