Pierce™ Antibody Biotinylation Kit for IP
Pierce™ Antibody Biotinylation Kit for IP
Thermo Scientific™
Pierce™ Antibody Biotinylation Kit for IP
Label primary antibodies with biotin for use in agarose or magnetic bead immunoprecipitation experiments using the Thermo Scientific™ Pierce™ Antibody Biotinylation Kit for IP.
Have Questions?
Catalog NumberQuantity
904078 Reactions
Catalog number 90407
Price (USD)
358.70
Special offer
Online exclusive
Ends: 15-Aug-2025
422.00
Save 63.30 (15%)
Each
Add to cart
Quantity:
8 Reactions
Request bulk or custom format
Price (USD)
358.70
Special offer
Online exclusive
Ends: 15-Aug-2025
422.00
Save 63.30 (15%)
Each
Add to cart
The Pierce Antibody Biotinylation Kit reagents have been optimized and validated to biotin label antibodies for IP and co-IP reactions. Determining the optimal number of biotins to attach to the target molecule is one of the major challenges of biotinylation. For IP and co-IP applications, too many biotins result in reduced affinity for the target antigen, while too few biotins result in antibody leaching upon elution of the target antigen. The biotin labeling procedure in the Pierce Antibody Biotinylation Kit for IP has been developed to address this challenge.
The Thermo Scientific™ Pierce™ Antibody Biotinylation Kit for IP provides biotinylation reagents designed specifically for the labeling of primary antibodies used in immunoprecipitation applications.
  • Optimized for IP - reagents and protocols developed specifically to address antibody labeling for immunoprecipitation applications
  • Easy to use - kit configuration specifically optimized to accommodate 50 to 200μg of antibody in a 100μL reaction; desalting columns are provided to remove excess biotinylation reagent after labeling
  • Amine reactive - reacts with primary amines (-NH2) of antibodies
  • Enhanced solubility - pegylation improves water solubility of the biotinylated antibody and prevents aggregation upon storage
  • Reduced steric hindrance - longer spacer arm (29 angstroms) minimize steric hindrance when binding to avidin molecules
The kit contains sufficient reagents to label 50 to 200μg of antibody in 100μL reaction volumes for eight samples. The NHS-PEG4-biotin labeling reagent contains an amine-reactive N-hydroxysuccinimide ester (NHS) group and a water-soluble PEG4 spacer for optimal labeling and is provided in easy-to-use, single-use microtubes. Both the labeling efficiency of the biotinylation reagent and binding affinity of the labeled antibody have been validated using mouse monoclonal (IgG1, IgG2), rabbit polyconal, and rabbit monoclonal antibodies. Zeba™ Desalting Spin Columns are provided for easy and efficient removal of salts and excess biotin.
Applications:
  • IP and co-IP
  • IP and co-IP for mass spectrometry applications
  • Antibody attachment to streptavidin agarose and magnetic supports
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Labeling Scale100 μg
Product LinePierce™
Product TypeAntibody Biotinylation Kit
Quantity8 Reactions
Labeling TargetAntibodies
Label or DyeBiotin
Unit SizeEach
Contents & Storage
EZ-Link NHS-PEG4-Biotin, 8 x 0.5 mg
Zeba Spin Desalting Columns (7K MWCO), 8 x 0.5 mL
20X Phosphate Buffered Saline, 15 mL
Store in refrigerator (2–8°C).
Frequently asked questions (FAQs)
I'd like to use the Pierce Antibody Biotinylation Kit for IP (Cat. No. 90407). Is it okay if my antibody storage buffer contains sodium azide?
Make sure that the final concentration of sodium azide in the reaction volume does not exceed 0.02%. The Zeba Spin Desalting Columns provided in the Pierce Antibody Biotinylation Kit for IP can be used to remove sodium azide from the storage solution.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link