Search Thermo Fisher Scientific
Catalog Number | Description |
---|---|
910199 | Standard All-in-One pH Buffer Kit |
810199 | ROSS All-in-One pH Buffer Kit |
916099 | Small Bottle All-in-One pH Buffer Kit |
Thermo Scientific™ Orion™ All-in-One™ pH buffer kits include pH 4.01, 7.00 and 10.01 buffers that are color-coded for easy identification as well as storage solution to help ensure your pH electrode is properly maintained. Using high-quality pH buffers to calibrate your pH electrode and pH meter is the best way to ensure your pH measurements are accurate and reproducible. Orion pH buffers are NIST traceable and manufactured under ISO 9001 quality standards.