Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Ensure fresh, ready-to-use standards for pH calibration with Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches. Buffers are color coded for ease of use. All Orion pH buffers are traceable to NIST™ standard reference materials.
Chemical Name or Material | pH Buffer, individual use pouches |
Boiling Point | 100°C |
Color | Light Blue |
Description | pH 10.01 Buffer |
For Use With (Application) | pH calibration |
For Use With (Equipment) | pH Meter and Electrode, single-use |
pH | 10.01 |
Physical Form | Liquid |
Quantity | 10 x 15 mL |
Solution Type | Buffer |
Formulation | Water, Sodium Bicarbonate, Methylparaben, FD & C Blue #1, Sodium Carbonate |
Packaging | Pouch |
Traceability to NIST | Yes |
Solubility Information | Soluble |
Unit Size | Pack of 10 |
Chemical Name or Material | pH Buffer, individual use pouches |
Boiling Point | 100°C |
Color | Light Red |
Description | pH 4.01 Buffer |
For Use With (Application) | pH calibration |
For Use With (Equipment) | pH Meter and Electrode, single-use |
pH | 4.01 |
Physical Form | Liquid |
Quantity | 10 x 15 mL |
Solution Type | Buffer |
Formulation | Water, Potassium Hydroxide, Amaranth Red No. 27, Potassium Hydrogen Phthalate |
Packaging | Pouch |
Traceability to NIST | Yes |
Solubility Information | Soluble |
Unit Size | Case of 10 |
Chemical Name or Material | pH Buffer, individual use pouches |
Boiling Point | 100°C |
Color | Light Red |
Description | pH 4.01 Buffer |
For Use With (Application) | pH calibration |
For Use With (Equipment) | pH Meter and Electrode, single-use |
pH | 4.01 |
Physical Form | Liquid |
Quantity | 25 x 15 mL |
Solution Type | Buffer |
Formulation | Water, Potassium Hydroxide, Amaranth Red No. 27, Potassium Hydrogen Phthalate |
Packaging | Pouch |
Traceability to NIST | Yes |
Solubility Information | Soluble |
Unit Size | Pack of 25 |
Chemical Name or Material | pH Buffer, individual use pouches |
Boiling Point | 100°C |
Color | Light Yellow |
Description | pH 7.00 Buffer |
For Use With (Application) | pH calibration |
For Use With (Equipment) | pH Meter and Electrode, single-use |
pH | 7 |
Physical Form | Liquid |
Quantity | 10 x 15 mL |
Solution Type | Buffer |
Formulation | Water, Dibasic, Potassium Dihydrogen Phosphate, 5-Bromo-5-Nitro-1,3-Dioxane, FD & C Yellow #5, Sodium Phosphate |
Packaging | Pouch |
Traceability to NIST | Yes |
Solubility Information | Soluble |
Unit Size | Pack of 10 |
Chemical Name or Material | pH Buffer, individual use pouches |
Boiling Point | 100°C |
Color | Light Yellow |
Description | pH 7.00 Buffer |
For Use With (Application) | pH calibration |
For Use With (Equipment) | pH Meter and Electrode, single-use |
pH | 7 |
Physical Form | Liquid |
Quantity | 25 x 15 mL |
Solution Type | Buffer |
Formulation | Water, Dibasic, Potassium Dihydrogen Phosphate, 5-Bromo-5-Nitro-1,3-Dioxane, FD & C Yellow #5, Sodium Phosphate |
Packaging | Pouch |
Traceability to NIST | Yes |
Solubility Information | Soluble |
Unit Size | Pack of 25 |
Chemical Name or Material | pH Buffer, individual use pouches |
Boiling Point | 100°C |
Color | Light Blue |
Description | pH 10.01 Buffer |
For Use With (Application) | pH calibration |
For Use With (Equipment) | pH Meter and Electrode, single-use |
pH | 10.01 |
Physical Form | Liquid |
Quantity | 25 x 15 mL |
Solution Type | Buffer |
Formulation | Water, Sodium Bicarbonate, Methylparaben, FD & C Blue #1, Sodium Carbonate |
Packaging | Pouch |
Traceability to NIST | Yes |
Solubility Information | Soluble |
Unit Size | Case of 25 |
Catalog Number | Specifications | Unit Size | pH | Packaging | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
911010 | Pack of 10 | 10.01 | Pouch | 10 x 15 mL | 39.73 Online price 46.20 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
910410-WA | Case of 10 | 4.01 | Pouch | 10 x 15 mL | 39.73 Online price 46.20 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
910425 | Pack of 25 | 4.01 | Pouch | 25 x 15 mL | 70.20 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
910710 | Pack of 10 | 7 | Pouch | 10 x 15 mL | 39.73 Online price 46.20 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
910725 | Pack of 25 | 7 | Pouch | 25 x 15 mL | 70.20 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
911025-WA | Case of 25 | 10.01 | Pouch | 25 x 15 mL | 70.20 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Note:
The following part numbers have been changed due to a recent system upgrade.
Old Part Number: 910410
New Part Number: 910410-WA
Old Part Number: 911025
New Part Number: 911025-WA
Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.
Warranty: Out of box