Perform applications using the Thermo Scientific™ Orion™ 9156DJWP Double Junction pH Electrode, featuring a durable epoxy body and low-maintenance gel.
Have Questions?
Catalog number 9156DJWP
Price (USD)/ Each
236.84
Special Offer
Online exclusive
Ends: 31-Dec-2024
277.00
Save 40.16 (15%)
-
Add to cart
Description:
Double Junction pH Electrode
Price (USD)/ Each
236.84
Special Offer
Online exclusive
Ends: 31-Dec-2024
277.00
Save 40.16 (15%)
Add to cart
Orion™ 9156DJWP Double Junction pH Electrode
Catalog number9156DJWP
Price (USD)/ Each
236.84
Special Offer
Online exclusive
Ends: 31-Dec-2024
277.00
Save 40.16 (15%)
-
Add to cart
Perform applications using the Thermo Scientific™ Orion™ 9156DJWP Double Junction pH Electrode, featuring a durable epoxy body and low-maintenance gel.
Ordering Information:
9156DJWP
Specifications
DescriptionDouble Junction pH Electrode
JunctionWick
MaterialEpoxy Body
pH Range0 to 14
Temperature (Metric) Operating0°C to 60°C
TypeDouble Junction pH Electrode
Internal ReferenceAg/AgCl Double Junction
Precision0.02
Storage RequirementsStore in Orion pH electrode storage solution 910001