Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
Thermo Scientific™
Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FilteredSterilityFit for PurposeUnit Size
917-021130 μLFilteredSterileSterile & PCR ReadyCase of 4800
915-02120 μLFilteredSterileSterile & PCR ReadyCase of 4800
915-26120 μLNon-filteredSterileSterileCase of 4800
915-261PK
also known as 915-261
20 μLNon-filteredSterileSterilePack of 960
915-26220 μLNon-filteredNon-sterileCertified QualityCase of 4800
915-262PK
also known as 915-262
20 μLNon-filteredNon-sterileCertified QualityPack of 960
917-261200 μLNon-filteredSterileSterileCase of 4800
917-261PK
also known as 917-261
200 μLNon-filteredSterileSterilePack of 960
917-262200 μLNon-filteredNon-sterileCertified QualityCase of 4800
917-262PK
also known as 917-262
200 μLNon-filteredNon-sterileCertified QualityPack of 960
Catalog number 917-021
Price (USD)/ Case of 4800
-
Volume (Metric):
130 μL
Filtered:
Filtered
Sterility:
Sterile
Fit for Purpose:
Sterile & PCR Ready
Unit Size:
Case of 4800
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
Volume (Metric)130 μL
FilteredFiltered
SterilitySterile
ColorClear
FormatRack
Tip StyleExtended Length
For Use With (Equipment)Beckman Coulter Liquid Handling Systems
Product LineBeckman, Beckman
Fit for PurposeSterile & PCR Ready
Unit SizeCase of 4800