Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) | Filtered | Sterility | Fit for Purpose | Unit Size |
---|---|---|---|---|---|
921-262 | 100 μL | Non-filtered | Non-sterile | Certified Quality | Case of 4800 |
920-021 | 50 μL | Filtered | Sterile | Sterile & PCR Ready | Case of 4800 |
920-261 | 50 μL | Non-filtered | Sterile | Sterile | Case of 4800 |
920-262 | 50 μL | Non-filtered | Non-sterile | Certified Quality | Case of 4800 |
921-021 | 80 μL | Filtered | Sterile | Sterile & PCR Ready | Case of 4800 |
921-021PK also known as 921-021 | 80 μL | Filtered | Sterile | Sterile & PCR Ready | Pack of 960 |
921-261 | 100 μL | Non-filtered | Sterile | Sterile | Case of 4800 |
921-261PK also known as 921-261 | 100 μL | Non-filtered | Sterile | Sterile | Pack of 960 |
921-262PK also known as 921-262 | 100 μL | Non-filtered | Non-sterile | Certified Quality | Pack of 960 |
923-021 | 150 μL | Filtered | Sterile | Sterile & PCR Ready | Case of 4800 |
923-021PK also known as 923-021 | 150 μL | Filtered | Sterile | Sterile & PCR Ready | Pack of 960 |
923-261 | 200 μL | Non-filtered | Sterile | Sterile | Case of 4800 |
923-262 | 200 μL | Non-filtered | Non-sterile | Certified Quality | Case of 4800 |
923-262PK also known as 923-262 | 200 μL | Non-filtered | Non-sterile | Certified Quality | Pack of 960 |
Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.