Search Thermo Fisher Scientific
Increase pipetting effectiveness with these automation tips, achieving accurate and precise performance due to superior tip straightness and a low coefficient of variation.
Catalog Number | Volume (Metric) | Autoclavable | Filtered | Fit for Purpose | Unit Size |
---|---|---|---|---|---|
927-021 | 40 μL | Not Autoclavable | Filtered | Sterile & PCR Ready | Case of 4800 |
925-021 | 10 μL | Not Autoclavable | Filtered | Sterile & PCR Ready | Case of 4800 |
925-261 | 20 μL | Not Autoclavable | Non-filtered | Sterile | Case of 4800 |
925-261PK also known as 925-261 | 20 μL | Not Autoclavable | Non-filtered | Sterile | Pack of 960 |
925-262 | 20 μL | Autoclavable | Non-filtered | Certified Quality | Case of 4800 |
927-021PK also known as 927-021 | 40 μL | Not Autoclavable | Filtered | Sterile & PCR Ready | Pack of 960 |
927-261 | 50 μL | Not Autoclavable | Non-filtered | Sterile | Case of 4800 |
927-261PK also known as 927-261 | 50 μL | Not Autoclavable | Non-filtered | Sterile | Pack of 960 |
927-262 | 50 μL | Autoclavable | Non-filtered | Certified Quality | Case of 4800 |
927-262PK also known as 927-262 | 50 μL | Autoclavable | Non-filtered | Certified Quality | Pack of 960 |
929-021 | 180 μL | Not Autoclavable | Filtered | Sterile & PCR Ready | Case of 4800 |
929-021PK also known as 929-021 | 180 μL | Not Autoclavable | Filtered | Sterile & PCR Ready | Pack of 960 |
929-261 | 235 μL | Not Autoclavable | Non-filtered | Sterile | Case of 4800 |
929-261PK also known as 929-261 | 235 μL | Not Autoclavable | Non-filtered | Sterile | Pack of 960 |
929-262 | 235 μL | Autoclavable | Non-filtered | Certified Quality | Case of 4800 |
929-262PK also known as 929-262 | 235 μL | Autoclavable | Non-filtered | Certified Quality | Pack of 960 |
931-021 | 20 μL | Not Autoclavable | Filtered | Sterile & PCR Ready | Case of 19,200 |
927-261-05 | 50 μL | Not Autoclavable | Non-filtered | Sterile | Case of 4800 |
Thermo Scientific™ Automation Tips for Perkin Elmer™/Packard™ Robotic Liquid Handling Systems with Varispan™ Dispense Arms, which undergo a rigorous 15-point quality control program to ensure consistency. This leads to accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.