Search Thermo Fisher Scientific
Provide safe and convenient storage for manual and electronic pipettes using Thermo Scientific™ Finnpipette™ Stands. Available options include options for single stand, F-stand, multi-stand, multichannel and carousel.
Product Type | Pipette Stand |
Description | Pipette Stand, gray |
Color | Gray |
Holds | Manual Pipettes (6) |
Positions | 6 |
Unit Size | Each |
Product Type | Pipette Stand |
Description | F-Stand, Linear Stand, white |
Holds | Manual Pipettes (6) |
Positions | 6 |
Unit Size | Each |
Product Type | Pipette Stand |
Description | Finnpipette Shelf Hanger |
Holds | Manual Pipette (1) |
Unit Size | Each |
Product Type | Pipette Stand |
Holds | 3 |
Positions | 3 |
Unit Size | Pack of 3 |
Product Type | Carousel Stand Adapter |
Description | Finnpipette Novus Adapter for Carousel Stand |
Color | White |
Holds | 1 |
Positions | 1 |
Unit Size | Each |
Product Type | Pipette Stand |
Description | Pipette Mini-Stand, gray |
Color | Gray |
Holds | Manual Pipettes (3) |
Positions | 3 |
Unit Size | Each |
Product Type | Pipette Stand |
Color | White |
Holds | Pipettes (6 Manual or 3 Electronic) |
Unit Size | Each |
Product Type | Pipette Stand |
Color | White |
Holds | Electronic Pipette (1) |
Unit Size | Each |
Product Type | Pipette Stand |
Description | Multichannel Single Pipette Stand |
Holds | Multichannel Manual Pipette (1) |
Positions | 1 |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Holds | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
9420290 | Each | Pipette Stand, gray | Manual Pipettes (6) | 101.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
9420400 | Each | F-Stand, Linear Stand, white | Manual Pipettes (6) | 88.04 Online price 111.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2206040 | Each | Finnpipette Shelf Hanger | Manual Pipette (1) | Request A Quote | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2209480 | Pack of 3 | 3 | Request A Quote | - | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2209490 | Each | Finnpipette Novus Adapter for Carousel Stand | 1 | Request A Quote | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
9420320 | Each | Pipette Mini-Stand, gray | Manual Pipettes (3) | 79.80 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
9420340 | Each | Pipettes (6 Manual or 3 Electronic) | 356.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
9420360 | Each | Electronic Pipette (1) | 127.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
9420390 | Each | Multichannel Single Pipette Stand | Multichannel Manual Pipette (1) | 41.60 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Finnpipette F-Stand
Finnpipette Multichannel Stand
Finnpipette Carousel Stand
Finnpipette Novus Stand