Graphite Furnace Cuvettes for Atomic Absorption (AA) Spectrometers
Graphite Furnace Cuvettes for Atomic Absorption (AA) Spectrometers
Actual product may vary
Graphite Furnace Cuvettes for Atomic Absorption (AA) Spectrometers
Graphite Furnace Cuvettes for Atomic Absorption (AA) Spectrometers
Graphite Furnace Cuvettes for Atomic Absorption (AA) Spectrometers
Graphite Furnace Cuvettes for Atomic Absorption (AA) Spectrometers
Graphite Furnace Cuvettes for Atomic Absorption (AA) Spectrometers
Thermo Scientific™
Graphite Furnace Cuvettes for Atomic Absorption (AA) Spectrometers
Improve your AAS graphite furnace performance using Thermo Scientific™ application-specific graphite cuvettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
942339395071Pyro-Coated Cuvettes
942339395041Extended Lifetime Cuvettes
942349020101Omega Cuvette with Platform
Catalog number 942339395071
Price (USD)
917.00
Each
Add to cart
Type:
Pyro-Coated Cuvettes
Price (USD)
917.00
Each
Add to cart
Ask our AI about this Product
Discover different types of graphite furnace cuvettes suitable for any analytical application using the Thermo Scientific™ GFS33/GFS35/GFS35Z Graphite Furnace and Autosampler modules
Improve instrument performance with the right selection of graphite cuvettesWhile ridged cuvettes are supplied as standard with Thermo Scientific GFS33/GFS35/GFS35Z Graphite Furnaces, additional application-specific cuvettes improve performance for more complex applications:
  • Extended Lifetime Cuvettes (ELCs) can be used longer and provide reproducible performance over several thousand firings
  • Ridged Cuvettes (in pyrolytically-coated and uncoated forms) help maintain difficult samples at the optimum temperature for efficient atomization
  • Omega Cuvettes with integral platforms provide the most interference-free conditions (often in combination with matrix modification) for samples with complex matrices
Specifications
For Use With (Equipment)GFS33 or GFS35 Furnaces
TypePyro-Coated Cuvettes
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection