Thermo Scientific™
ID100 Consumables Kit for iCE™ 3000 Series AAS instruments
Catalog number: 942345003201
Have Questions?
Thermo Scientific™
ID100 Consumables Kit for iCE™ 3000 Series AAS instruments
Catalog number: 942345003201
Have Questions?
Get everything you need in one kit. The Thermo Scientific™ ID100 Consumables Kit for the Thermo Scientific™ iCE™ 3000 Series AAS instruments ensures quick and easy connectivity for the Thermo Scientific ID100 Flame Autodilutor. For fast, in-line dilution with high accuracy and reduced pump wear-and-tear, the ID100 Flame Autodilutor is an excellent add-on to the cost effective Thermo Scientific 3000 Series Flame AA spectrometers and the advanced Thermo Scientific™ iCAP™ Q Series ICP-MS systems.
Catalog Number
942345003201
Unit Size
Each
Description
ID100 Consumables Kit
Price (USD)
Full specifications
Quantity
TypeConsumables Kit
DescriptionID100 Consumables Kit
For Use With (Equipment)iCE 3000 Series Flame AA Spectrometers
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)Quantity
942345003201Full specifications
EachID100 Consumables Kit
TypeConsumables Kit
DescriptionID100 Consumables Kit
For Use With (Equipment)iCE 3000 Series Flame AA Spectrometers
Unit SizeEach
Showing 1 of 1
Get everything you need in one kit. The Thermo Scientific™ ID100 Consumables Kit for the Thermo Scientific™ iCE™ 3000 Series AAS instruments ensures quick and easy connectivity for the Thermo Scientific ID100 Flame Autodilutor. For fast, in-line dilution with high accuracy and reduced pump wear-and-tear, the ID100 Flame Autodilutor is an excellent add-on to the cost effective Thermo Scientific 3000 Series Flame AA spectrometers and the advanced Thermo Scientific™ iCAP™ Q Series ICP-MS systems.