Thermo Scientific™
EC100 Electrically Heated Cell System
Catalog number: 942346020001
Thermo Scientific™
EC100 Electrically Heated Cell System
Catalog number: 942346020001
Switch from a flame-heated atomization cell to the Thermo Scientific™ EC100 Electrically Heated Cell System and gain the option of safe, unattended, high-sensitivity vapor generation Atomic Absorption analysis, plus the benefit of reduced operating costs.
Catalog Number
942346020001
Unit Size
Each
Description
EC100 Electrically Heated Cell System
Price (EUR)
Full specifications
TypeHeated Cell System
DescriptionEC100 Electrically Heated Cell System
For Use With (Equipment)iCE 3000 Series Flame AA Spectrometers
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (EUR)
942346020001Full specifications
EachEC100 Electrically Heated Cell SystemRequest A Quote
TypeHeated Cell System
DescriptionEC100 Electrically Heated Cell System
For Use With (Equipment)iCE 3000 Series Flame AA Spectrometers
Unit SizeEach
Showing 1 of 1
Improved Analysis Capability with Vapor AA
  • Optimized atomization temperature for higher sensitivity and improved precision in comparison to flame heating
  • Simple and rapid T-cell replacement minimizes down time
  • Suitable for all hydride forming elements
  • For use with the Thermo Scientific™ VP100 Vapor Generation accessory and our Thermo Scientific™ iCE™ 3000 Series AA Spectrometers