Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | ClipTip 1000, Filtered, Sterile, Rack |
Volume (Metric) | 1000 μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Length (English) | 3.74 in. |
Length (Metric) | 9.5 cm |
Format | Rack |
Tip Style | Low Retention |
Quantity | 8 Racks (96 Tips/Rack) |
Color Code | Blue |
For Use With (Equipment) | Thermo Scientific E1-ClipTip, Thermo Scientific F1-ClipTip |
Product Line | ClipTip |
Fit for Purpose | Forensic Ready |
Unit Size | Pack of 768 |
Description | ClipTip 12.5 Ext, Filtered, Sterile, Rack |
Volume (Metric) | 12.5 μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Length (English) | 1.61 in. |
Length (Metric) | 4.1 cm |
Format | Rack |
Tip Style | Extended Length |
Quantity | 10 Racks (96 Tips/Rack) |
Color Code | Pink |
For Use With (Equipment) | Thermo Scientific E1-ClipTip, Thermo Scientific F1-ClipTip |
Product Line | ClipTip |
Fit for Purpose | Forensic Ready |
Unit Size | Pack of 960 |
Description | ClipTip 20, Filtered, Sterile, Rack |
Volume (Metric) | 20 μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Length (English) | 1.8 in. |
Length (Metric) | 4.6 cm |
Format | Rack |
Tip Style | Low Retention |
Quantity | 10 Racks (96 Tips/Rack) |
Color Code | Pink |
For Use With (Equipment) | Thermo Scientific E1-ClipTip, Thermo Scientific F1-ClipTip |
Product Line | ClipTip |
Fit for Purpose | Forensic Ready |
Unit Size | Pack of 960 |
Description | ClipTip 200, Filtered, Sterile, Rack |
Volume (Metric) | 200 μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Length (English) | 2.2 in. |
Length (Metric) | 5.6 cm |
Format | Rack |
Tip Style | Low Retention |
Quantity | 10 Racks (96 Tips/Rack) |
Color Code | Yellow |
For Use With (Equipment) | Thermo Scientific E1-ClipTip, Thermo Scientific F1-ClipTip |
Product Line | ClipTip |
Fit for Purpose | Forensic Ready |
Unit Size | Pack of 960 |
Catalog Number | Specifications | Unit Size | Description | Volume (Metric) | Color Code | Fit for Purpose | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
94420713-HID | Pack of 768 | ClipTip 1000, Filtered, Sterile, Rack | 1000 μL | Blue | Forensic Ready | 153.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
94420063-HID | Pack of 960 | ClipTip 12.5 Ext, Filtered, Sterile, Rack | 12.5 μL | Pink | Forensic Ready | 136.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
94420213-HID | Pack of 960 | ClipTip 20, Filtered, Sterile, Rack | 20 μL | Pink | Forensic Ready | 138.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
94420313-HID | Pack of 960 | ClipTip 200, Filtered, Sterile, Rack | 200 μL | Yellow | Forensic Ready | 138.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Key Features
• ClipTip pipette tips feature 'clips' that lock tips in place and form a complete seal
• Each time ClipTip pipette tips are on, they are sealed and will not loosen regardless of application pressure
• ClipTip pipette tips are clipped securely on the F1-ClipTip and E1-ClipTip pipettes with a light touch eliminating the frustration of banging tips on your pipette
• Filtered ClipTip pipette tips are ideal for sensitive assays, such as PCR, and with critical samples to prevent cross-contaminations
Low retention
Low retention technology minimizes liquid retention, and enable maximium sample recovery. Available in ClipTip 20-1250 96-format tips.
Proven quality
Certified free of RNase, DNase, DNA, ATP and endotoxin contamination
Environmentally responsible
ClipTip reload system uses recycled components and 30% less plastic
Versatile Tip Range
• 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250 μL
• 384-format volumes: 12.5, 30 and 125 μL
• Extended lenght tips available for volumes 12.5 and 300 μL allowing for better reach with specific labware consumables
Immediate traceability
Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert
Note:
Availability may vary by country