Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Achieve superior reversed phase oligonucleotide separations using the Thermo Scientific™ DNAPac™ RP HPLC columns. The unique chemistry is designed for analysis of oligonucleotides and double-stranded (ds) DNA/RNA fragments using LC-UV or LC-MS. The column chemistry provides excellent performance under a broad range of pH, temperature, and mobile phase compositions. In addition, the wide pore size of the resin provides excellent separation of large double-stranded nucleic acids up to 10k base pairs.
Column Format | Semi-preparative Guard Cartridge Holder |
Diameter (Metric) | 21.2 mm |
Length (Metric) | 10 mm |
Quantity | Each |
Temperature | up to 110°C |
Product Line | DNAPac |
Type | Preparative Column |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 3 mm |
Length (Metric) | 100 mm |
Max. Pressure | 4000 psi (275 bar) |
Particle Size | 4 μm |
pH | 0 to 14 |
Pore Size | Proprietary Wide Pore |
Quantity | 1 Ea. |
Temperature | <110°C |
Stationary Phase | DVB (Divinylbenzene) Polymer |
Product Line | DNAPac |
Packing Material | DVB (Divinylbenzene) Polymer |
Column Type | Reversed Phase |
Phase | Reversed Phase |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 3 mm |
Length (Metric) | 50 mm |
Max. Pressure | 4000 psi (275 bar) |
Particle Size | 4 μm |
pH | 0 to 14 |
Pore Size | Proprietary Wide Pore |
Quantity | 1 Ea. |
Temperature | <110°C |
Stationary Phase | DVB (Divinylbenzene) Polymer |
Product Line | DNAPac |
Packing Material | DVB (Divinylbenzene) Polymer |
Column Type | Reversed Phase |
Phase | Reversed Phase |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Semi-preparative Column |
Diameter (Metric) | 10 mm |
Length (Metric) | 150 mm |
Max. Pressure | 4000 psi (275 bar) |
Particle Size | 4 μm |
pH | 0 to 14 |
Pore Size | Proprietary Wide Pore |
Temperature | 110°C |
Stationary Phase | DVB (Divinylbenzene) Polymer |
Product Line | DNAPac |
Packing Material | DVB (Divinylbenzene) Polymer |
Column Type | Reversed Phase |
Phase | Reversed Phase |
Unit Size | Each |
Column Format | Guard Cartridge |
Diameter (Metric) | 3 mm |
Length (Metric) | 10 mm |
Max. Pressure | 4000 psi (275 bar) |
Particle Size | 4 μm |
pH | 0 to 14 |
Pore Size | Proprietary Wide Pore |
Quantity | 2 Pk. |
Temperature | <110°C |
Stationary Phase | DVB (Divinylbenzene) Polymer |
Product Line | DNAPac |
Packing Material | DVB (Divinylbenzene) Polymer |
Column Type | Reversed Phase |
Phase | Reversed Phase |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 2.1 mm |
Length (Metric) | 100 mm |
Max. Pressure | 4000 psi (275 bar) |
Particle Size | 4 μm |
pH | 0 to 14 |
Pore Size | Proprietary Wide Pore |
Quantity | 1 Ea. |
Temperature | <110°C |
Stationary Phase | DVB (Divinylbenzene) Polymer |
Product Line | DNAPac |
Packing Material | DVB (Divinylbenzene) Polymer |
Column Type | Reversed Phase |
Phase | Reversed Phase |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 2.1 mm |
Length (Metric) | 50 mm |
Max. Pressure | 4000 psi (275 bar) |
Particle Size | 4 μm |
pH | 0 to 14 |
Pore Size | Proprietary Wide Pore |
Quantity | 1 Ea. |
Temperature | <110°C |
Stationary Phase | DVB (Divinylbenzene) Polymer |
Product Line | DNAPac |
Packing Material | DVB (Divinylbenzene) Polymer |
Column Type | Reversed Phase |
Phase | Reversed Phase |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Guard Cartridge |
Diameter (Metric) | 2.1 mm |
Length (Metric) | 10 mm |
Max. Pressure | 4000 psi (275 bar) |
Particle Size | 4 μm |
pH | 0 to 14 |
Pore Size | Proprietary Wide Pore |
Quantity | 2 Pk. |
Temperature | <110°C |
Stationary Phase | DVB (Divinylbenzene) Polymer |
Product Line | DNAPac |
Packing Material | DVB (Divinylbenzene) Polymer |
Column Type | Reversed Phase |
Phase | Reversed Phase |
Technique | Bio-LC |
Unit Size | Each |
Diameter (Metric) | 2.1 mm |
For Use With | HPLC |
Length (Metric) | 250 mm |
Max. Pressure | 4000 psi (275 bar) |
Particle Shape | Spherical |
Particle Size | 4 μm |
pH | 1 to 14 |
Pore Size | Proprietary Wide Pore |
Temperature | <110°C |
Packing Material | DVB |
Column Type | Analytical |
Endcapped | No |
Phase | Reversed Phase |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Semi-preparative Guard Cartridge |
Diameter (Metric) | 21.2 mm |
Flow Rate | 4 to 8 mL/min |
Length (Metric) | 10 mm |
Max. Pressure | 4000 psi (275 bar) |
pH | 0 to 14 |
Quantity | 2 PK. |
Stationary Phase | Proprietary |
Product Line | DNAPac |
Packing Material | 4 µm polymer Resin |
Column Type | Reversed Phase |
Phase | Reversed Phase |
Material | DVB (Divinylbenzene) Polymer |
Type | Semi-preparative |
Technique | Reversed Phase HPLC |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Diameter (Metric) | Length (Metric) | Price (USD) | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
950-00 | Each | 21.2 mm | 10 mm | Request A Quote | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088919 | Each | 3 mm | 100 mm | 1,011.21 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088920 | Each | 3 mm | 50 mm | 866.59 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SP6998 | Each | 10 mm | 150 mm | Request A Quote | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088921 | Each | 3 mm | 10 mm | 261.26 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088923 | Each | 2.1 mm | 100 mm | 1,001.88 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088924 | Each | 2.1 mm | 50 mm | 866.59 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088925 | Each | 2.1 mm | 10 mm | 261.26 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
303324 | Each | 2.1 mm | 250 mm | 1,527.90 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
080922-0121232 | Each | 21.2 mm | 10 mm | Request A Quote | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The DNAPac RP column is based on spherical wide-pore 4μm polymer resin. The phase is highly stable to extremes of pH (0-14), and temperature (5-110°C), and operates with mobile phases compatible with direct mass spectrometry. The column may be operated in both HPLC and UHPLC instruments, producing high resolution at high throughput for oligonucleotides and resolves both small and very large dsDNA segments. DNAPac RP reversed phase (RP) column is specifically designed for separation of oligonucleotides and DNA/RNA fragments. The stationary phase is fully compatible with mass spectrometry friendly organic solvents and ion-pair reagents such as triethylamine (TEA) and hexylamine (HA). The wide pore size of polymer resin enables efficient separation of large nucleic acids with low carry-over.
Columns designed for reversed phase oligonucleotide LC and LC/MS separations