Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.
Breakapart | Breakable Strips |
Binding Property | Hydrophobic |
Certified | No |
Format | Strip Plate |
Description | High optical quality for reliable results |
Detection Method | Colorimetric |
Packaging | 25 Pk., 50 Cases |
Binding Type | Adsorption |
Color | Clear |
Product Type | Microplate Strip |
Material | Polystyrene |
Volume (Metric) Well | 400 μL |
No. of Wells | 96 |
Volume (Metric) Working | 350 μL |
Surface Treatment | Universal Binding |
Well Shape | Round, Flat Bottom (F-well) |
For Use With (Application) | For adsorption of hydrophobic molecules |
No. per Case | 50 |
No. per Pack | 50 |
Plate Blocking | None |
Target Molecule | Lipids |
Unit Size | Case of 50 |
Breakapart | Breakable Strips (LockWell) |
Binding Property | Hydrophobic |
Certified | No |
Format | Strip Plate |
Lid | Without Lid |
Description | High optical quality for reliable results |
Detection Method | Colorimetric |
Array | 8 x 12 |
Packaging | Inner packs of 5 |
Color | Clear |
Product Type | Strip Plate |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Sterility | Non-sterile |
Surface Treatment | PolySorp |
Well Shape | Round, C-bottom |
For Use With (Application) | For adsorption of hydrophobic molecules |
No. per Case | 60 |
No. per Pack | 5 |
Plate Blocking | None |
Quantity | 60/Cs. |
Target Molecule | Lipids |
Well Design | C8 |
Unit Size | Case of 60 |
Breakapart | Breakable Strips (LockWell) |
Binding Property | Hydrophilic |
Certified | Yes |
Format | Strip Plate |
Lid | Without Lid |
Description | High optical quality for reliable results |
Detection Method | Colorimetric |
Array | 8 x 12 |
Packaging | 10 Pk. |
Color | Clear |
Product Type | Strip Plate |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Sterility | Non-sterile |
Surface Treatment | MaxiSorp |
Well Shape | Round, C-bottom |
For Use With (Application) | For antibody sandwich assays |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Quantity | 60/Cs. |
Target Molecule | Antibodies (Glycoproteins) |
Well Design | C8 |
Unit Size | Case of 60 |
Breakapart | Breakable Strips |
Binding Property | Hydrophobic |
Certified | No |
Format | Strip Plate |
Description | High optical quality for reliable results |
Detection Method | Colorimetric |
Packaging | 10 Pk. |
Color | Clear |
Product Type | Frame |
Material | Polystyrene |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Surface Treatment | PolySorp |
Well Shape | Round |
For Use With (Application) | For adsorption of hydrophobic molecules |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Target Molecule | Lipids |
Unit Size | Each |
Breakapart | Breakable Strips |
Binding Property | Very Hydrophilic |
Certified | No |
Format | Strip Plate |
Lid | Without Lid |
Description | High optical quality for reliable results |
Detection Method | Colorimetric |
Array | 8 x 12 |
Packaging | 10 Pk. |
Color | Clear |
Product Type | Frame |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Surface Treatment | MultiSorp |
Well Shape | Round, C-bottom |
For Use With (Application) | For glycans and water soluble proteins |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Target Molecule | Glycans |
Unit Size | Case of 60 |
Breakapart | Breakable Strips |
Binding Property | Hydrophilic |
Certified | Yes |
Format | Strip Plate |
Description | High optical quality for reliable results |
Detection Method | Colorimetric |
Packaging | 10 Pk., 60 Cases |
Color | Clear |
Product Type | Microplate Strip |
Material | Polystyrene |
Volume (Metric) Well | 320 μL |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Surface Treatment | MaxiSorp |
Well Shape | Round |
For Use With (Application) | For antibody sandwich assays |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Target Molecule | Antibodies (Glycoproteins) |
Unit Size | Case of 60 |
Binding Property | Hydrophobic |
Certified | No |
Format | Strip Plate |
Description | High optical quality for reliable results |
Detection Method | Colorimetric |
Array | 8 x 12 |
Packaging | 10 Pk., 60 Cases |
Color | Clear |
Product Type | Microplate Strip |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Surface Treatment | PolySorp |
Well Shape | Round, C-bottom |
For Use With (Application) | For adsorption of hydrophobic molecules |
No. per Case | 50 |
No. per Pack | 5 |
Plate Blocking | None |
Quantity | 50/Cs. |
Target Molecule | Lipids |
Unit Size | Case of 60 |
Breakapart | Breakable Strips |
Binding Property | Hydrophilic |
Certified | Yes |
Format | Strip Plate |
Lid | Without Lid |
Description | High optical quality for reliable results |
Detection Method | Colorimetric |
Array | 8 x 12 |
Packaging | 10 Pk., 60 Cases |
Binding Type | Adsorption |
Color | Clear |
Product Type | Strip Plate |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Surface Treatment | MaxiSorp |
Well Shape | Round, C-bottom |
For Use With (Application) | For antibody sandwich assays |
No. per Case | 60 |
No. per Pack | 5 |
Plate Blocking | None |
Quantity | 60/Cs. |
Target Molecule | Antibodies (Glycoproteins) |
Well Design | C8 |
Unit Size | Case of 60 |
Catalog Number | Specifications | Unit Size | Surface Treatment | Well Shape | No. per Pack | Product Type | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
95029390 | Case of 50 | Universal Binding | Round, Flat Bottom (F-well) | 50 | Microplate Strip | 422.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
446442 | Case of 60 | PolySorp | Round, C-bottom | 5 | Strip Plate | 528.30 Online price 587.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
446469 | Case of 60 | MaxiSorp | Round, C-bottom | 10 | Strip Plate | 550.80 Online price 612.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
446477 | Each | PolySorp | Round | 10 | Frame | Request A Quote | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
446490 | Case of 60 | MultiSorp | Round, C-bottom | 10 | Frame | 418.00 Online price 612.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
446639 | Case of 60 | MaxiSorp | Round | 10 | Microplate Strip | Request A Quote | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
473539 | Case of 60 | PolySorp | Round, C-bottom | 5 | Microplate Strip | 930.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
473768 | Case of 60 | MaxiSorp | Round, C-bottom | 5 | Strip Plate | 635.00 Online price 930.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|