3UV Lamp (230V, 50Hz)
3UV Lamp (230V, 50Hz)
Thermo Scientific™

3UV Lamp (230V, 50Hz)

Thermo Scientific Pierce UV Lamps are compact, multifunctional ultraviolet lamps with 252nm, 302nm and 365nm dial-selectable wavelength settings especially forRead more
Have Questions?
Catalog number 95035
Price (USD)/ 1 unit
898.00
-
Add to cart
Price (USD)/ 1 unit
898.00
Add to cart
3UV Lamp (230V, 50Hz)
Catalog number95035
Price (USD)/ 1 unit
898.00
-
Add to cart
Thermo Scientific Pierce UV Lamps are compact, multifunctional ultraviolet lamps with 252nm, 302nm and 365nm dial-selectable wavelength settings especially for use with photoactivatable crosslinkers and UV-crosslinking methods. This 230V, 50Hz lamp is designed for grounded outlets in continental Europe. Adaptor required for the UK.

Featuresof Pierce UV Lamps:

Multifunctional—three lamps (254nm, 302nm, 365nm) selected by a turn of the dial
Efficient—unique reflector design provides maximum UV for all applications
Lightweight and ergonomic—designed for hand-held or stationary use
Small footprint—15.5' Length x 3' Width x 4.5' Depth (394mm x 76mm x 114mm)
Durable—lamp features aluminum construction and scratch-resistant finish
Versatile—8-watt lamps are ideal for a wide area of illumination, such as culture plates
Validated for crosslinking—our researchers have validated this lamp for use in crosslinking applications with diazirine and phenyl azide photoactivatable crosslinkers

These 3UV lamp from UVP is a compact ultraviolet lamp for longwave, midrange and shortwave UV applications. The lamp's innovative design allows you to dial-in the required wavelength with just the turn of a knob. The lamp is compatible with our extensive line of photo-reactive crosslinking and label-transfer reagents, including the Photo-reactive Amino Acids for discovering in-cell protein interactions.

Applications:
• Photo-reactive crosslinkers
• Photo-reactive biotinylation
Photo-reactive biotin label-transfer reagents
• Photo-reactive amino acids (

Related Products
3UV Lamp (115V, 60Hz)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeTriple Wavelength UV Lamp
Light SourceUV
Voltage230 V
Wattage8 W
Wavelength365/302/254 nm
DescriptionPierce 3UV Lamp, 230 Volts, 50 Hz (Europe)
Unit Size1 unit