Nunc™ 96-Well Polypropylene DeepWell™ Sample Processing & Storage Plates with Shared-Wall Technology
Nunc™ 96-Well Polypropylene DeepWell™ Sample Processing & Storage Plates with Shared-Wall Technology
Nunc™ 96-Well Polypropylene DeepWell™ Sample Processing & Storage Plates with Shared-Wall Technology
Nunc™ 96-Well Polypropylene DeepWell™ Sample Processing & Storage Plates with Shared-Wall Technology
Nunc™ 96-Well Polypropylene DeepWell™ Sample Processing & Storage Plates with Shared-Wall Technology
Thermo Scientific™

Nunc™ 96-Well Polypropylene DeepWell™ Sample Processing & Storage Plates with Shared-Wall Technology

Optimize storage capacity and improve mixing using these DeepWell™ storage plates featuring shared-wall technology. Shared-Wall reduces the height of the plate, increases the diameter of the well for sample extraction.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityWell ShapeVolume (Metric) Well
95040462SterileSquare, Conical Bottom (V-well)2000 μL
260251SterileRound1300 μL
260252Non-sterileRound1300 μL
278743SterileRound2000 μL
278752Non-sterileRound2000 μL
95040452Non-sterileSquare, Conical Bottom (V-well)2000 μL
Catalog number 95040462
Price (USD)
437.01
Online Exclusive
497.00
Save 59.99 (12%)
Case of 50
Add to cart
Sterility:
Sterile
Well Shape:
Square, Conical Bottom (V-well)
Volume (Metric) Well:
2000 μL
Price (USD)
437.01
Online Exclusive
497.00
Save 59.99 (12%)
Case of 50
Add to cart
Ask our AI about this Product

Optimize storage capacity and improve mixing using Thermo Scientific™ Nunc™ 1.3 and 2.0mL DeepWell™ Plates with Shared-Wall Technology. Ideal for sample collection, storage, combinatorial chemistry and library applications, and resistant to most chemicals, solvents and alcohols used in combinatorial chemistry.

The large well diameters offer ideal access and excellent mixing characteristics, and the U shaped well enhances compatibility with the magnetic bead extraction. The V shaped well bottom ensures low dead-volume and high sample recovery during extraction

All Thermo Scientific plates are manufactured in our cleanroom facility or clean environment to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.

Features:

  • Ideal as a collection plate for Nunc Filter Plates
  • Storage plate for compounds, samples or biomolecules
  • Round-bottom wells reduce liquid retention
  • Round well enhances mixing
  • Automated molecular diagnostic workflows
  • Magnetic Bead Extraction compatible
  • Ideal for Kitting

Working volumes:

  • 260251 and 260252 50-1000 μL/well
  • 278743 and 278752 50-1900 μL/well

Recommended for:

Sample collection; storage; combinatorial chemistry; drug library applications; bacterial and yeast growth; collection plates for Nunc filter plates; NGS and genomic library prep

Compatible Sealing: Adhesive and Heat seals
Hazardous: No
Max Centrifugal Force: 2000 g

All plates are ANSI format for compatibility with automated systems.

To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.

Specifications
AutoclavableAutoclavable
Binding TypeLow binding PP
Certifications/ComplianceCertified RNase and DNase Free. ANSI Compliant.
Dnase FreeYes
Description96-well Polypropylene Sample Processing & Storage Plates
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
LidWithout Lid
No. per Case50
No. per Pack1
PackagingIndividually wrapped
Plate SizeANSI/ SLAS Standard
Product Type96-well plate
Quantity50/Cs.
RNase FreeYes
Regulatory StatusFor Research Use Only. Not for use in diagnostic procedures.
Shelf Life5 years
Volume (Metric) Working1800 μL
Array8 x 12
BarcodeCustom Barcoding Available
ColorNatural
Fit for PurposeCertified Quality
FormatStandard
MaterialPolypropylene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentNon-treated
Volume (Metric) Well2000 μL
Well ShapeSquare, Conical Bottom (V-well)
Unit SizeCase of 50
web-banner-9-8-22-278x123

Fit for Purpose designations

Gain confidence that the products you buy are ideally suited to your application. 

Learn more ›

nalgene-certificate-of-compliance

Certificates of Compliance

Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Nalgene and Nunc products.
Start your request  

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link