951-021 Full specifications
Pack of 960 250 μL Filtered 96 Tips/Tray, 10 Trays/Case Sterile & PCR Ready Request A Quote - - Volume (Metric) 250 μL Filtered Filtered Sterility Sterile Color Clear Format Rack Tip Style Automation Packaging 96 Tips/Tray, 10 Trays/Case For Use With (Equipment) Universal Fit Pipettor, Qiagen Product Line Qiagen, Qiagen Fit for Purpose Sterile & PCR Ready Unit Size Pack of 960
951-253 Full specifications
Case of 4,800 300 μL Non-Filtered 96 Tips/Tray, 50 Trays/Case Sterile Contact Us
- - Volume (Metric) 300 μL Filtered Non-Filtered Sterility Sterile Color Black Format Rack Tip Style Conductive Packaging 96 Tips/Tray, 50 Trays/Case For Use With (Equipment) Universal Fit Pipettor, Qiagen Product Line Qiagen Fit for Purpose Sterile Unit Size Case of 4,800
951-261 Full specifications
Case of 960 300 μL Non-Filtered 96 Tips/Tray, 10 Trays/Case Sterile 100.00 Online price 172.00
- Volume (Metric) 300 μL Filtered Non-Filtered Sterility Sterile Color Clear Format Rack Tip Style Automation Packaging 96 Tips/Tray, 10 Trays/Case For Use With (Equipment) Universal Fit Pipettor, Qiagen Product Line Qiagen Fit for Purpose Sterile Unit Size Case of 960
951-262 Full specifications
Pack of 960 300 μL Non-Filtered 96 Tips/Tray, 10 Trays/Case Certified Quality Request A Quote - - Volume (Metric) 300 μL Filtered Non-Filtered Sterility Non-sterile Color Clear Format Rack Tip Style Automation Packaging 96 Tips/Tray, 10 Trays/Case For Use With (Equipment) Universal Fit Pipettor, Qiagen Product Line Qiagen Fit for Purpose Certified Quality Unit Size Pack of 960
953-021 Full specifications
Case of 960 1000 μL Filtered 96 Tips/Tray, 10 Trays/Case Sterile & PCR Ready 174.65 Online price 229.00
- Volume (Metric) 1000 μL Filtered Filtered Sterility Sterile Color Clear Format Rack Tip Style Automation Packaging 96 Tips/Tray, 10 Trays/Case For Use With (Equipment) Universal Fit Pipettor, Qiagen Product Line Qiagen, Qiagen Fit for Purpose Sterile & PCR Ready Unit Size Case of 960
953-253 Full specifications
Case of 4800 1100 μL Non-Filtered 96 Tips/Tray, 50 Trays/Case Sterile 736.65 Online price 869.00
- Volume (Metric) 1100 μL Filtered Non-Filtered Sterility Non-sterile Color Clear Format Rack Tip Style Conductive Packaging 96 Tips/Tray, 50 Trays/Case For Use With (Equipment) Universal Fit Pipettor, Qiagen Product Line Qiagen Fit for Purpose Sterile Unit Size Case of 4800
953-261 Full specifications
Case of 960 1100 μL Non-Filtered 96 Tips/Tray, 10 Trays/Case Sterile 137.65 Online price 174.00
- Volume (Metric) 1100 μL Filtered Non-Filtered Sterility Sterile Color Clear Format Rack Tip Style Automation Packaging 96 Tips/Tray, 10 Trays/Case For Use With (Equipment) Universal Fit Pipettor, Qiagen Product Line Qiagen Fit for Purpose Sterile Unit Size Case of 960
953-262 Full specifications
Pack of 960 1100 μL Non-Filtered 96 Tips/Tray, 10 Trays/Case Certified Quality Request A Quote - - Volume (Metric) 1100 μL Filtered Non-Filtered Sterility Non-sterile Color Clear Format Rack Tip Style Automation Packaging 96 Tips/Tray, 10 Trays/Case For Use With (Equipment) Universal Fit Pipettor, Qiagen Product Line Qiagen Fit for Purpose Certified Quality Unit Size Pack of 960