AcroMetrix™ Herpes Simplex Virus Controls (RUO)
Product Image
Thermo Scientific™
AcroMetrix™ Herpes Simplex Virus Controls (RUO)
Assess the performance of molecular diagnostic test procedures for human herpes viruses such as HSV-1 and 2 with these controls, which are for Research Use Only.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
951300AcroMetrix HHV Negative Control
951301AcroMetrix HSV-1 Low Control
951302AcroMetrix HSV-2 Low Control
954502AcroMetrix HSV-2 High CSF Control
954501AcroMetrix HSV-1 High CSF Control
Catalog number 951300
Price (USD)
-
Request A Quote
Description:
AcroMetrix HHV Negative Control
Assess the performance of nucleic acid test procedures for the quantitative and qualitative determination of Human Herpes Viruses (HHV) with the Thermo Scientific™ AcroMetrix™ Herpes Simplex Virus Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionAcroMetrix HHV Negative Control
TypeHHV Control
Packaging5 x 0.5mL
Quantity5 x 0.5 mL
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link