AcroMetrix SARS-CoV-2 Control (Full-process, RUO)
AcroMetrix SARS-CoV-2 Control (Full-process, RUO)
AcroMetrix SARS-CoV-2 Control (Full-process, RUO)
AcroMetrix SARS-CoV-2 Control (Full-process, RUO)
Thermo Scientific™

AcroMetrix SARS-CoV-2 Control (Full-process, RUO)

Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19 using the Thermo Scientific™ AcroMetrix™ SARS-CoV-2 Control (RUO)
Have Questions?
KatalognummerMenge
9545171 Kit
Katalognummer 954517
Preis (EUR)
-
Angebot anfordern
Menge:
1 Kit
Ask our AI about this Product
Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19 using the Thermo Scientific™ AcroMetrix SARS-CoV-2 Control (RUO). This control mimics a patient sample and can be used as a complete assay control starting from extraction to amplification and finally detection of COVID-19.
  • Inactivated SARS-CoV-2 in proprietary viral transport media
  • Full process control mimics a patient sample
  • Qualitative positive control
  • Kit contains 5 vials per kit x 1.5 mL per vial
Ausschliesslich zu Forschungszwecken. Nicht zugelassen für diagnostische Verfahren.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BeschreibungAcroMetrix SARS-CoV-2 Control (RUO)
VerpackungOne kit contains 5 vials per kit x 1.5 mL per vial
Menge1 Kit
Unit SizeEach
Inhalt und Lagerung
-20 °C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Dokumente und Downloads

Zertifikate

Sicherheitsdatenblätter

Geben Sie die Katalognummer, den Namen oder Link frei.

1x1 image pixel for data collection