1x1 image pixel for data collection
AcroMetrix SARS-CoV-2 Control (In-process, RUO)
AcroMetrix SARS-CoV-2 Control (In-process, RUO)
Thermo Scientific™
AcroMetrix SARS-CoV-2 Control (In-process, RUO)
Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19 using the Thermo Scientific™ AcroMetrix™ Coronavirus 2019 (COVID-19) RNA Control (RUO).
Have Questions?
Catalog NumberQuantity
954519Each
Catalog number 954519
Price (USD)
500.00
Each
Add to cart
Quantity:
Each
Price (USD)
500.00
Each
Add to cart
Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19 using the Thermo Scientific™ AcroMetrix™ Coronavirus 2019 (COVID-19) RNA Control (RUO). This is a positive control kit containing two concentrations: a Low Positive and Ultra-Low Positive concentration to avoid false-negative results.
  • Full length genomic RNA from SARS-CoV-2
  • Kit contains one Low Positive control and one Ultra-low Positive control
  • Quantitated using Bio-Rad Droplet Digital™ PCR (ddPCR™)
For Research Use Only.
Available in select countries only.
Specifications
DescriptionAcroMetrix Coronavirus 2019 (COVID-19) RNA Control (RUO)
PackagingOne kit contains 2 vials per kit x 0.02mL per vial
QuantityEach
Unit SizeEach
Contents & Storage
-20°C or lower
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link