TaqMan™ SARS-CoV-2, Flu A/B, RSV RNA Control
TaqMan™ SARS-CoV-2, Flu A/B, RSV RNA Control
TaqMan™ SARS-CoV-2, Flu A/B, RSV RNA Control
TaqMan™ SARS-CoV-2, Flu A/B, RSV RNA Control
TaqMan™ SARS-CoV-2, Flu A/B, RSV RNA Control
TaqMan™ SARS-CoV-2, Flu A/B, RSV RNA Control
TaqMan™ SARS-CoV-2, Flu A/B, RSV RNA Control
Applied Biosystems™
TaqMan™ SARS-CoV-2, Flu A/B, RSV RNA Control
The TaqMan SARS-CoV-2, Flu A/B, RSV RNA Control contains targets specific to the SARS-CoV-2, influenza A/B, and RSV genomic regionsRead more
Have Questions?
Catalog NumberQuantity
95612610 x 10 μL
Catalog number 956126
Price (USD)
-
Request A Quote
Quantity:
10 x 10 μL
Ask our AI about this Product
The TaqMan SARS-CoV-2, Flu A/B, RSV RNA Control contains targets specific to the SARS-CoV-2, influenza A/B, and RSV genomic regions targeted by the TaqMan SARS-CoV-2, FluA/B RSV RT-PCR Assay Kit. It is recommended for use with this assay as a positive control to ensure reaction setup and reagent integrity when interpreting test results.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use WithTaqMan™ SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit
FormatTube(s)
Product LineTaqMan™
Quantity10 x 10 μL
Shipping ConditionDry Ice
Unit Size10 x 10 µL
Contents & Storage
Store at ≤-70°C.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
266912Certificate of AnalysisFeb 12, 2025956126, 956126LSG
279786_0425Certificate of AnalysisFeb 12, 2025956126, 956126LSG
257313Certificate of AnalysisFeb 12, 2025956126, 956126LSG
275786_011025Certificate of AnalysisFeb 12, 2025956126, 956126LSG
260014Certificate of AnalysisFeb 12, 2025956126, 956126LSG
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection