Nunc™ Microscope Slides
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ Microscope Slides
Thermo Scientific™

Nunc™ Microscope Slides

Exchange glass slides with Thermo Scientific™ Nunc™ Microscope Slides, which offer greater cell attachment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial
960004Polystyrene
160005Permanox
Catalog number 960004
Price (USD)
417.00
Case of 100
Add to cart
Material:
Polystyrene
Price (USD)
417.00
Case of 100
Add to cart

Achieve similar cell attachment and growth as standard cell culture flasks and dishes with the Thermo Scientific™ Nunc™ Plastic Microscope Slides. Available in either polystyrene or Permanox plastics.

  • Surface treatment of the plastic slides ensures optimal cell attachment and growth, similar to that in the regular cell culture vessels
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
CoatingCell-Culture Treated
DescriptionPolystyrene microscope slide
Dimensions (L x W)75 x 25 mm
No. per Case100
No. per Pack20
SterilitySterile
Length (Metric)75 mm
MaterialPolystyrene
Width (Metric)25 mm
Unit SizeCase of 100

Frequently asked questions (FAQs)

Which fixation reagents can I use with Permanox plastic?

We have a Technical Bulletin (link below) describing fixation reagents that are compatible with Permanox plastic: https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/D19581.pdf

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link