AcroMetrix NAT Blood Screening Controls
Product Image
Thermo Scientific™
AcroMetrix NAT Blood Screening Controls
Assess performance of NAT assays that test human plasma from donations of whole blood and blood components with these controls, which are for In Vitro Diagnostic Use.
Have Questions?
Catalog NumberDescriptionDetectable AnalytesPackaging
960500AcroMetrix WNVRNA ControlWest Nile Virus (WNV)10 x 1.4mL
Catalog number 960500
Price (USD)
-
Request A Quote
Description:
AcroMetrix WNVRNA Control
Detectable Analytes:
West Nile Virus (WNV)
Packaging:
10 x 1.4mL
Ask our AI about this Product
Estimate the precision and reproducibility of NAT assays that are designed to detect viral nucleic acids in human plasma from donations of whole blood and blood components for transfusion with the Thermo Scientific™ AcroMetrix NAT Blood Screening Controls. These controls mimic naturally occurring plasma specimens and can help detect potential systematic deviations of tests for the qualitative determination of viral nucleic acids. The controls are for In Vitro Diagnostic Use.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionAcroMetrix WNVRNA Control
Detectable AnalytesWest Nile Virus (WNV)
Sample TypePlasma
TypeControl
Packaging10 x 1.4mL
Quantity10 x 1.4 mL
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link