Nitric acid, 69.0-70.0%, NF, GMP, J.T.Baker™
Nitric acid, 69.0-70.0%, NF, GMP, J.T.Baker™
Avantor™

Nitric acid, 69.0-70.0%, NF, GMP, J.T.Baker™

This product is GMP Manufactured and meets N.F. Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Clear glass bottle
Clear glass bottle, PVC-coated
Catalog number 9607-04
Price (USD)
-
Request A Quote
Quantity:
2.5 L
Packaging:
Clear glass bottle
Ask our AI about this Product
This product is GMP Manufactured and meets N.F. Requirements.
This product is GMP Manufactured and meets N.F. Requirements.
Specifications
Chemical Name or MaterialNitric acid
ColorColorless to slightly yellow
CAS7697-37-2
Melting Point-42°C
Boiling Point120.5°C (1013 hPa)
PackagingClear glass bottle
Quantity2.5 L
Molecular FormulaHNO3
SynonymAqua Fortis, Azotic Acid
Molecular Weight (g/mol)63.01
Percent Purity69.0-70.0%
GradeNF
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection