Search Thermo Fisher Scientific
Measure cadmium ions in aqueous solutions quickly, simply and economically with the Thermo Scientific™ Orion™ Cadmium Electrodes.
9648BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
Connector Types | Waterproof BNC |
Description | Model 9448BN; Orion Cadmium Combination |
Junction | Free-flowing Sure-Flow junction assures stable, drift-free potentials |
For Use With (Equipment) | ISE Meters |
Type | Combination Ion Selective Electrode |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
9648BNWP | Each | 1,143.00 Online price 1,270.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|