Thermo Scientific™
Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
Catalog number: 967-262PK
Have Questions?
Thermo Scientific™
Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
Catalog number: 967-262PK
Have Questions?
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System are available in non-sterile, sterile and sterile filtered tips. Each pipet tip undergoes a rigorous 15-point quality control program to ensure consistency, thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
Catalog Number
967-262PK
also known as 967-262
Unit Size
Pack of 960
Volume (Metric)
1000 μL
Filtered
Non-Filtered
Sterility
Non-sterile
Fit for Purpose
Certified Quality
Price (USD)
Availability
-
Full specifications
Quantity
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeCertified Quality
Unit SizePack of 960
Catalog Number
Unit Size
Case of 3840
Volume (Metric)
50 μL
Filtered
Filtered
Sterility
Sterile
Fit for Purpose
Sterile & PCR Ready
Price (USD)
Availability
-
Full specifications
Quantity
Volume (Metric)50 μL
FilteredFiltered
SterilitySterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile & PCR Ready
Unit SizeCase of 3840
Catalog Number
965-021PK
also known as 965-021
Unit Size
Pack of 960
Volume (Metric)
50 μL
Filtered
Filtered
Sterility
Sterile
Fit for Purpose
Sterile & PCR Ready
Price (USD)
Availability
-
Full specifications
Quantity
Volume (Metric)50 μL
FilteredFiltered
SterilitySterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile & PCR Ready
Unit SizePack of 960
Catalog Number
Unit Size
Case of 3840
Volume (Metric)
50 μL
Filtered
Non-Filtered
Sterility
Sterile
Fit for Purpose
Sterile
Price (USD)
Availability
-
Full specifications
Quantity
Volume (Metric)50 μL
FilteredNon-Filtered
SterilitySterile
ColorClear
FormatRack
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile
Unit SizeCase of 3840
Catalog Number
965-261PK
also known as 965-261
Unit Size
Pack of 960
Volume (Metric)
50 μL
Filtered
Non-Filtered
Sterility
Sterile
Fit for Purpose
Sterile
Price (USD)
Availability
-
Full specifications
Quantity
Volume (Metric)50 μL
FilteredNon-Filtered
SterilitySterile
ColorClear
FormatRack
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile
Unit SizePack of 960
Catalog Number
Unit Size
Case of 3840
Volume (Metric)
50 μL
Filtered
Non-Filtered
Sterility
Non-sterile
Fit for Purpose
Certified Quality
Price (USD)
Availability
-
Full specifications
Quantity
Volume (Metric)50 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeCertified Quality
Unit SizeCase of 3840
Catalog Number
965-262PK
also known as 965-262
Unit Size
Pack of 960
Volume (Metric)
50 μL
Filtered
Non-Filtered
Sterility
Non-sterile
Fit for Purpose
Certified Quality
Price (USD)
Availability
-
Full specifications
Quantity
Volume (Metric)50 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeCertified Quality
Unit SizePack of 960
Catalog Number
Unit Size
Case of 3840
Volume (Metric)
300 μL
Filtered
Filtered
Sterility
Sterile
Fit for Purpose
Sterile & PCR Ready
Price (USD)
Availability
-
Full specifications
Quantity
Volume (Metric)300 μL
FilteredFiltered
SterilitySterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile & PCR Ready
Unit SizeCase of 3840
Catalog Number
966-021PK
also known as 966-021
Unit Size
Pack of 960
Volume (Metric)
300 μL
Filtered
Filtered
Sterility
Sterile
Fit for Purpose
Sterile & PCR Ready
Price (USD)
Availability
-
Full specifications
Quantity
Volume (Metric)300 μL
FilteredFiltered
SterilitySterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile & PCR Ready
Unit SizePack of 960
Catalog Number
Unit Size
Case of 3840
Volume (Metric)
300 μL
Filtered
Non-Filtered
Sterility
Sterile
Fit for Purpose
Sterile
Price (USD)
Availability
-
Full specifications
Quantity
Volume (Metric)300 μL
FilteredNon-Filtered
SterilitySterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile
Unit SizeCase of 3840
Showing 10 of 18Show more
Catalog NumberSpecificationsUnit SizeVolume (Metric)FilteredSterilityFit for PurposePrice (USD)AvailabilityQuantity
967-262PK
also known as 967-262
Full specifications
Pack of 9601000 μLNon-FilteredNon-sterileCertified Quality
-
Volume (Metric)1000 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeCertified Quality
Unit SizePack of 960
965-021Full specifications
Case of 384050 μLFilteredSterileSterile & PCR Ready
-
Volume (Metric)50 μL
FilteredFiltered
SterilitySterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile & PCR Ready
Unit SizeCase of 3840
965-021PK
also known as 965-021
Full specifications
Pack of 96050 μLFilteredSterileSterile & PCR Ready
-
Volume (Metric)50 μL
FilteredFiltered
SterilitySterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile & PCR Ready
Unit SizePack of 960
965-261Full specifications
Case of 384050 μLNon-FilteredSterileSterile
-
Volume (Metric)50 μL
FilteredNon-Filtered
SterilitySterile
ColorClear
FormatRack
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile
Unit SizeCase of 3840
965-261PK
also known as 965-261
Full specifications
Pack of 96050 μLNon-FilteredSterileSterile
-
Volume (Metric)50 μL
FilteredNon-Filtered
SterilitySterile
ColorClear
FormatRack
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile
Unit SizePack of 960
965-262Full specifications
Case of 384050 μLNon-FilteredNon-sterileCertified Quality
-
Volume (Metric)50 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeCertified Quality
Unit SizeCase of 3840
965-262PK
also known as 965-262
Full specifications
Pack of 96050 μLNon-FilteredNon-sterileCertified Quality
-
Volume (Metric)50 μL
FilteredNon-Filtered
SterilityNon-sterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeCertified Quality
Unit SizePack of 960
966-021Full specifications
Case of 3840300 μLFilteredSterileSterile & PCR Ready
-
Volume (Metric)300 μL
FilteredFiltered
SterilitySterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile & PCR Ready
Unit SizeCase of 3840
966-021PK
also known as 966-021
Full specifications
Pack of 960300 μLFilteredSterileSterile & PCR Ready
-
Volume (Metric)300 μL
FilteredFiltered
SterilitySterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile & PCR Ready
Unit SizePack of 960
966-261Full specifications
Case of 3840300 μLNon-FilteredSterileSterile
-
Volume (Metric)300 μL
FilteredNon-Filtered
SterilitySterile
ColorClear
FormatRack
Tip StyleAutomation
For Use With (Equipment)Eppendorf epMotion
Product LineEppendorf epMotion
Fit for PurposeSterile
Unit SizeCase of 3840
Showing 10 of 18Show more
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Warranty:A five (5)-year shelf life from the date of manufacture is recommended.