AcroMetrix™ HIV-1/HCV/HBV Controls (IVD)
Product Image
Thermo Scientific™
AcroMetrix™ HIV-1/HCV/HBV Controls (IVD)
Assess the performance of molecular assays that detect human immunodeficiency virus type 1, Hepatitis B and C with bulk controls, which are for In Vitro Diagnostic Use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevel
967151High
967150Low
Catalog number 967151
Price (USD)
-
Request A Quote
Level:
High
Ask our AI about this Product
Assess the performance of molecular assays that detect human immunodeficiency virus type 1, Hepatitis C and Hepatitis B in patient samples with the Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Controls (IVD). Provided in 50mL bulk format and composed of multiple analytes, the AcroMetrix HIV-1/HCV/HBV Controls provide end-user workflow flexibility and cost-efficiency. Manufactured in accordance with ISO 17511, these controls are designed to yield consistent results from lot-to-lot and are ideal for monitoring assay performance over time. The controls are for In Vitro Diagnostic Use.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Detectable AnalytesHIV-1, HCV, HBV
TypeHIV-1/HCV/HBV Control
LevelHigh
Packaging50mL
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
256362Certificate of AnalysisJan 29, 2025967150
254002Certificate of AnalysisJan 29, 2025967151
264832_033125Certificate of AnalysisJan 28, 2025967151
279491_033126Certificate of AnalysisJan 25, 2025967150
286454_053126Certificate of AnalysisJan 25, 2025967151
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection