Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™
KingFisher™ Systems Consumables
Catalog number: 97002070
Have Questions?
Thermo Scientific™
KingFisher™ Systems Consumables
Catalog number: 97002070
Have Questions?
Optimize performance using Thermo Scientific™ KingFisher™ Systems Consumables, specifically designed for use with the KingFisher System and ideal for magnetic particle processing because of their low binding affinity for biomolecules. The specially-designed, disposable tip combs and microplates, which are made of polypropylene, enable excellent recovery of magnetic beads.
Catalog Number
97002070
Unit Size
Case of 50
Description
KingFisher tip comb, 50 pcs/box
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionKingFisher tip comb, 50 pcs/box
For Use With (Application)Magnetic particle processing
For Use With (Equipment)KingFisher System
Quantity50/Cs.
Unit SizeCase of 50
Catalog Number
Unit Size
Case of 50
Description
KingFisher plate 100 μL, 50 pcs/box
Price (USD)
Availability
-
Full specifications
Quantity
Capacity100 μL
DescriptionKingFisher plate 100 μL, 50 pcs/box
For Use With (Application)Magnetic particle processing
For Use With (Equipment)KingFisher System
Quantity50/Cs.
Unit SizeCase of 50
Catalog Number
Unit Size
Case of 50
Description
KingFisher plate 200 μL, 50 pcs/box
Price (USD)
Availability
-
Full specifications
Quantity
Capacity200 μL
DescriptionKingFisher plate 200 μL, 50 pcs/box
For Use With (Application)Magnetic particle processing
For Use With (Equipment)KingFisher System
Quantity50/Cs.
Unit SizeCase of 50
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
97002070Full specifications
Case of 50KingFisher tip comb, 50 pcs/box
-
DescriptionKingFisher tip comb, 50 pcs/box
For Use With (Application)Magnetic particle processing
For Use With (Equipment)KingFisher System
Quantity50/Cs.
Unit SizeCase of 50
97002080Full specifications
Case of 50KingFisher plate 100 μL, 50 pcs/box
-
Capacity100 μL
DescriptionKingFisher plate 100 μL, 50 pcs/box
For Use With (Application)Magnetic particle processing
For Use With (Equipment)KingFisher System
Quantity50/Cs.
Unit SizeCase of 50
97002084Full specifications
Case of 50KingFisher plate 200 μL, 50 pcs/box
-
Capacity200 μL
DescriptionKingFisher plate 200 μL, 50 pcs/box
For Use With (Application)Magnetic particle processing
For Use With (Equipment)KingFisher System
Quantity50/Cs.
Unit SizeCase of 50
Showing 3 of 3
  • Specifically designed for use with the KingFisher System
  • Ideal for magnetic particle processing because of their low binding affinity for biomolecules
  • The specially-designed, disposable tip combs and microplates are made of polypropylene
  • This enables excellent recovery of magnetic beads
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.