KingFisher™ Plastics for 96 standard and PCR formats
KingFisher™ Plastics for 96 standard and PCR formats
Actual product may vary
Thermo Scientific™

KingFisher™ Plastics for 96 standard and PCR formats

Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Promo ImagePromotion
Promo code:P5780456
Gene Expression Promo
From RNA Isolation kits, reverse transcriptases to analysis of your gene with qPCR - upto 35% on all Gene Expression workflow productsLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Quantity
97002524BPromo ImageKingFisher Apex100/Cs.
97002560Promo ImageKingFisher Apex50/Cs.
97002540Promo ImageKingFisher Flex and Presto48/Cs.
97002524Promo ImageKingFisher Flex100/Cs.
97002514Promo ImageKingFisher Flex80/Cs.
97002540BPromo ImageKingFisher Apex48/Cs.
Catalog number 97002524B
Price (USD)
583.65
Online Exclusive
700.00
Save 116.35 (17%)
Case of 100
Add to cart
For Use With (Equipment):
KingFisher Apex
Quantity:
100/Cs.
Price (USD)
583.65
Online Exclusive
700.00
Save 116.35 (17%)
Case of 100
Add to cart
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionKingFisher 96 Tip Comb, Barcoded
For Use With (Equipment)KingFisher Apex
Quantity100/Cs.
Unit SizeCase of 100

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection