KingFisher™ Plastics for 96 standard and PCR formats
KingFisher™ Plastics for 96 standard and PCR formats
Thermo Scientific™

KingFisher™ Plastics for 96 standard and PCR formats

Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Catalog NumberFor Use With (Equipment)Quantity
97002540KingFisher Flex and Presto48/Cs.
97002560KingFisher Apex50/Cs.
97002524KingFisher Flex100/Cs.
97002514KingFisher Flex80/Cs.
97002524BKingFisher Apex100/Cs.
97002540BKingFisher Apex48/Cs.
Catalog number 97002540
Price (JPY)
28,700
Case of 48
Contact Us ›
For Use With (Equipment):
KingFisher Flex and Presto
Quantity:
48/Cs.
Ask our AI about this Product
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionKingFisher 96 microplate (200μL) (for Flex and Presto)
*Note: Only works with KingFisher™ Flex 96 Heating Block (Cat. No. 24075420). Not to be used with KingFisher™ Flex 96 Deep-Well Heating Block (Cat. No. 24075430).
For Use With (Equipment)KingFisher Flex and Presto
Quantity48/Cs.
Capacity96 Samples
Format96 Deep-well Plate
Unit SizeCase of 48
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
25152F0Certificate of ComplianceJun 26, 202597002540B
25169F0Certificate of ComplianceJun 26, 202597002540B
25097F0Certificate of ComplianceJun 26, 202597002540B
25148F0Certificate of ComplianceJun 26, 202597002540
25145F0Certificate of ComplianceJun 26, 202597002540
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection