Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | KingFisher 24 Deep-Well Tip Comb & Plate, Sterile, Barcoded |
For Use With (Equipment) | KingFisher Apex |
Quantity | 50/Cs. |
Unit Size | Case of 50 |
Format | 24 |
Capacity | 24 Samples |
Description | KingFisher Apex 24 Combi tip comb (for Apex only) |
Disposable | Single-use |
For Use With (Equipment) | For Use With KingFisher Apex only |
Quantity | 50/Cs. |
Unit Size | Case of 50 |
Format | 24 Deep-well Plate |
Capacity | 24 Samples |
Description | KingFisher 24 deep-well plate (for Duo Prime, Flex and Presto) |
For Use With (Equipment) | KingFisher Duo Prime, Flex and Presto |
Unit Size | Case of 50 |
Format | 24 Deep-well Plate |
Capacity | 24 Samples |
Description | KingFisher 24 deep-well plate, sterile, 50 pcs (for Duo Prime, Flex and Presto) |
For Use With (Equipment) | KingFisher Duo Prime, Flex and Presto |
Unit Size | Case of 50 |
Format | 24 Deep-well Comb |
Capacity | 24 Samples |
Description | KingFisher 24 deep-well tip comb and plate (for Flex and Presto) |
For Use With (Equipment) | KingFisher Flex and Presto |
Unit Size | Case of 50 |
Format | Plate and Comb |
Capacity | 24 Samples |
Description | KingFisher 24 deep-well, tip comb and plate, sterile (for Flex and Presto) |
For Use With (Equipment) | KingFisher Flex and Presto |
Unit Size | Case of 50 |
Format | Plate and Comb |
Description | KingFisher Duo Prime 6-tip comb and 24 deep-well plate (12 pieces of 24 deep-well plates, each including 4 tip combs)(for Duo Prime only) |
For Use With (Equipment) | For Use With KingFisher Duo Prime only |
Quantity | 48 Pc. |
Unit Size | Each |
Description | KingFisher 24 Deep-well Plate, Barcoded |
For Use With (Equipment) | KingFisher Apex |
Quantity | 50/Cs. |
Unit Size | Case of 50 |
Description | KingFisher 24 Deep-Well Tip Comb & Plate, Barcoded |
For Use With (Equipment) | KingFisher Apex |
Quantity | 50/Cs. |
Unit Size | Case of 50 |
Description | KingFisher 24 Deep-Well Plate, Sterile, Barcoded |
For Use With (Equipment) | KingFisher Apex |
Quantity | 50/Cs. |
Unit Size | Case of 50 |
Catalog Number | Specifications | Unit Size | Description | For Use With (Equipment) | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
97002620B | Case of 50 | KingFisher 24 Deep-Well Tip Comb & Plate, Sterile, Barcoded | KingFisher Apex | 1,004.65 Online price 1,112.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
97002580 | Case of 50 | KingFisher Apex 24 Combi tip comb (for Apex only) | For Use With KingFisher Apex only | 866.65 Online price 962.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
95040470 | Case of 50 | KingFisher 24 deep-well plate (for Duo Prime, Flex and Presto) | KingFisher Duo Prime, Flex and Presto | 320.65 Online price 356.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
95040480 | Case of 50 | KingFisher 24 deep-well plate, sterile, 50 pcs (for Duo Prime, Flex and Presto) | KingFisher Duo Prime, Flex and Presto | 398.65 Online price 442.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
97002610 | Case of 50 | KingFisher 24 deep-well tip comb and plate (for Flex and Presto) | KingFisher Flex and Presto | 943.65 Online price 1,048.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
97002620 | Case of 50 | KingFisher 24 deep-well, tip comb and plate, sterile (for Flex and Presto) | KingFisher Flex and Presto | 1,027.65 Online price 1,142.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
97003510 | Each | KingFisher Duo Prime 6-tip comb and 24 deep-well plate (12 pieces of 24 deep-well plates, each including 4 tip combs)(for Duo Prime only) | For Use With KingFisher Duo Prime only | 195.65 Online price 218.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
95040470B | Case of 50 | KingFisher 24 Deep-well Plate, Barcoded | KingFisher Apex | 328.65 Online price 365.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
97002610B | Case of 50 | KingFisher 24 Deep-Well Tip Comb & Plate, Barcoded | KingFisher Apex | 953.65 Online price 1,060.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
95040480B | Case of 50 | KingFisher 24 Deep-Well Plate, Sterile, Barcoded | KingFisher Apex | 386.65 Online price 429.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.