Rheodyne™ 9725 and 9725i Sample Injectors
Rheodyne™ 9725 and 9725i Sample Injectors
Thermo Scientific™
Rheodyne™ 9725 and 9725i Sample Injectors
Ensure precise HPLC injections and protect against pressure shock with biocompatible dual-mode Thermo Scientific™ Rheodyne™ 9725 and 9725i Sample Injections.
Have Questions?
Catalog number 9725I
Price (USD)/ Each
2,564.88
Add to cart
Description:
Dual mode; Continuous Flow; Position Sensing Switch
Price (USD)/ Each
2,564.88
Add to cart
Rheodyne™ 9725 and 9725i Sample Injectors
Catalog number9725I
Price (USD)/ Each
2,564.88
-
Add to cart
Use biocompatible Thermo Scientific™ Rheodyne™ 7725 and 7725i Sample Injectors to maintain continuous HPLC flow between the load and inject positions and to protect against pressure shock. The Make-Before-Break (MBB) design ensures uninterrupted flow during sample injection. Constructed of Thermo Scientific™ PEEK™, these dual-mode injectors can use partial filling for zero sample waste or complete filling for better reproducibility. Inject from 1μL to 5mL samples with high accuracy and precision. The 9725i model features a position-sensing switch for a reproducible start signal. The injectors include a 20μL sample loop.
  • Biocompatible PEEK™ construction
  • Make-Before-Break (MBB) design
  • Can use partial filling for zero sample waste or complete filling for better reproducibility
  • Inject 1μL to 5mL with high accuracy and precision
  • 9725i features a position sensing switch for a reproducible start signal
Includes:20μL sample loop
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
ModeDual Mode
Volume (Metric) Sample20 μL
DescriptionDual mode; Continuous Flow; Position Sensing Switch
For Use With (Equipment)HPLC
TypeContinuous Flow HPLC Sample Injector
Stator MaterialPEEK
Unit SizeEach
1x1 image pixel for data collection