Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Type | Quantity |
---|---|---|
984309 | Tartaric Acid System Reagent | 20 mL Vials |
984300 | Ethanol System Reagent | 20 mL Vials |
984302 | D to Fructose System Reagent | 20 mL Vials |
981842 | Wash Fluid | 20 mL Vials |
984304 | D to Glucose System Reagent | 20 mL Vials |
984332 | ECM pH 7 Standard Test | 20 mL Vials |
984353 | BC System Liquid | 20 mL Vials |
984354 | BC Diluent | 20 mL Vials |
984355 | BC Eluent | 20 mL Vials |
984764 | D to Glucose Reagent | 20 mL Vials |
984765 | Glycerol | 20 mL Vials |
984305 | Beta to Glucan System Reagent | 2 Reagents in 20 mL Vials |
984306 | D to Lactic Acid System Reagent | 20 mL Vials |
984307 | Potassium System Reagent | 1 Reagent in 20 mL Vial |
984308 | L to Lactic Acid System Reagent | 20 mL Vials |
984310 | L to Malic Acid System Reagent | 20 mL Vials |
984312 | Sucrose System Reagent | 20 mL Vials |
984314 | D to Glucose + D to Fructose System Reagent | 20 mL Vials |
984315 | Lipase System Reagent | 20 mL Vials |
984316 | Glycerol System Reagent | 20 mL Vials |
984317 | D to Glucose + D to Fructose + Sucrose System Reagent | 20 mL Vials |
984318 | Acetic Acid System Reagent | 20 mL Vials |
984319 | L to Asparagine/L to Aspartic Acid | 20 mL Vials |
984320 | Ammonia System Reagent | 20 mL Vials |
984321 | Urea System Reagent | 20 mL Vials |
984322 | D to Isocitric Acid System Reagent | 20 mL Vials |
984323 | Lactose System Reagent | 20 mL Vials |
984324 | Cholesterol in Food System Reagent | 20 mL Vials |
984326 | Total Iron System Reagent | 2 Reagents in 20 mL Vials, 1 Reagent Capsule |
984327 | Citric Acid System Reagent | 2 Working Reagents in 20 mL Vials |
984328 | Total Protein System Reagent | 1 Reagent in 20 mL Vial |
984342 | Alpha to Amino Nitrogen | 2 Reagents in 20 mL Vials |
984345 | Total Sulfite System Reagent | 2 Reagents in 20 mL Vials |
984346 | Total Polyphenol System Reagent | 2 Reagents in 20 mL Vials |
984347 | Acetaldehyde System Reagent | 2 Reagents in 20 mL Vials |
984348 | Oxalic Acid System Reagent | 2 Reagents in 20 mL Vials |
984349 | pH (Colorimetric) System Reagent | 1 Reagent in 20 mL Vial |
984356 | Acetic Acid System Reagent | 2 Working Reagents in 20 mL Vials |
984380 | Sugar Combination Standard Reagent | 3 x 3 mL |
984382 | Acid Combination Standard Reagent | 3 x 3 mL |
984383 | Beta to Glucan Standard Reagent | 1 x 50 mg |
984384 | Ethanol Standard Reagent | 10 x 1 mL |
984386 | Glycerol Standard Reagent | 3 x 3 mL |
984388 | Urea Standard Reagent | 3 x 3 mL |
984390 | Lactose Standard Reagent | 3 x 3 mL |
984391 | Cholesterol in Food Standard Reagent | 3 x 3 mL |
984393 | Oxalic Acid Standard Reagent | 3 x 3 mL |
984394 | NOPA Standard Reagent | 3 x 3 mL |
984396 | Acetaldehyde Standard | 3 x 5 mL |
984632 | Total Acids (Wine pH 7) | 1 buffer and 1 color reagent |
984633 | Total Acids (Juice pH 8) | 1 buffer and 1 color reagent |
984634 | Free Sulfite System Reagent | 2 Reagents in 20 mL Vials |
984635 | L to Ascorbic Acid System Reagent | 3 Working Reagents in 20 mL Vials |
984636 | L-glutamic Acid System Reagent | 20 mL Vials |
984710 | D to Gluconic Acid System Reagent | 2 Working Reagents in 20 mL Vials |
984373 | Alpha Amylase System Reagent | Reagent A: 6 x 20 mL Vials Reagent B: 2 x 5 mL Vials |
Find additional products for automated wet chemical analysis, including systems for beer, water and enzyme analysis as well as reagents for food and beverage or environmental and industrial analyses.
Share catalog number, name or link