Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Type | Quantity |
---|---|---|
984312 | Sucrose System Reagent | 20 mL Vials |
984300 | Ethanol System Reagent | 20 mL Vials |
984302 | D to Fructose System Reagent | 20 mL Vials |
981842 | Wash Fluid | 20 mL Vials |
984304 | D to Glucose System Reagent | 20 mL Vials |
984332 | ECM pH 7 Standard Test | 20 mL Vials |
984353 | BC System Liquid | 20 mL Vials |
984354 | BC Diluent | 20 mL Vials |
984355 | BC Eluent | 20 mL Vials |
984764 | D to Glucose Reagent | 20 mL Vials |
984765 | Glycerol | 20 mL Vials |
984305 | Beta to Glucan System Reagent | 2 Reagents in 20 mL Vials |
984306 | D to Lactic Acid System Reagent | 20 mL Vials |
984307 | Potassium System Reagent | 1 Reagent in 20 mL Vial |
984308 | L to Lactic Acid System Reagent | 20 mL Vials |
984309 | Tartaric Acid System Reagent | 20 mL Vials |
984310 | L to Malic Acid System Reagent | 20 mL Vials |
984314 | D to Glucose + D to Fructose System Reagent | 20 mL Vials |
984315 | Lipase System Reagent | 20 mL Vials |
984316 | Glycerol System Reagent | 20 mL Vials |
984317 | D to Glucose + D to Fructose + Sucrose System Reagent | 20 mL Vials |
984318 | Acetic Acid System Reagent | 20 mL Vials |
984319 | L to Asparagine/L to Aspartic Acid | 20 mL Vials |
984320 | Ammonia System Reagent | 20 mL Vials |
984321 | Urea System Reagent | 20 mL Vials |
984322 | D to Isocitric Acid System Reagent | 20 mL Vials |
984323 | Lactose System Reagent | 20 mL Vials |
984324 | Cholesterol in Food System Reagent | 20 mL Vials |
984326 | Total Iron System Reagent | 2 Reagents in 20 mL Vials, 1 Reagent Capsule |
984327 | Citric Acid System Reagent | 2 Working Reagents in 20 mL Vials |
984328 | Total Protein System Reagent | 1 Reagent in 20 mL Vial |
984342 | Alpha to Amino Nitrogen | 2 Reagents in 20 mL Vials |
984345 | Total Sulfite System Reagent | 2 Reagents in 20 mL Vials |
984346 | Total Polyphenol System Reagent | 2 Reagents in 20 mL Vials |
984347 | Acetaldehyde System Reagent | 2 Reagents in 20 mL Vials |
984348 | Oxalic Acid System Reagent | 2 Reagents in 20 mL Vials |
984349 | pH (Colorimetric) System Reagent | 1 Reagent in 20 mL Vial |
984356 | Acetic Acid System Reagent | 2 Working Reagents in 20 mL Vials |
984380 | Sugar Combination Standard Reagent | 3 x 3 mL |
984382 | Acid Combination Standard Reagent | 3 x 3 mL |
984383 | Beta to Glucan Standard Reagent | 1 x 50 mg |
984384 | Ethanol Standard Reagent | 10 x 1 mL |
984386 | Glycerol Standard Reagent | 3 x 3 mL |
984388 | Urea Standard Reagent | 3 x 3 mL |
984390 | Lactose Standard Reagent | 3 x 3 mL |
984391 | Cholesterol in Food Standard Reagent | 3 x 3 mL |
984393 | Oxalic Acid Standard Reagent | 3 x 3 mL |
984394 | NOPA Standard Reagent | 3 x 3 mL |
984396 | Acetaldehyde Standard | 3 x 5 mL |
984632 | Total Acids (Wine pH 7) | 1 buffer and 1 color reagent |
984633 | Total Acids (Juice pH 8) | 1 buffer and 1 color reagent |
984634 | Free Sulfite System Reagent | 2 Reagents in 20 mL Vials |
984635 | L to Ascorbic Acid System Reagent | 3 Working Reagents in 20 mL Vials |
984636 | L-glutamic Acid System Reagent | 20 mL Vials |
984710 | D to Gluconic Acid System Reagent | 2 Working Reagents in 20 mL Vials |
984373 | Alpha Amylase System Reagent | Reagent A: 6 x 20 mL Vials Reagent B: 2 x 5 mL Vials |
Share catalog number, name or link