Gallery™ System Reagents for Environmental and Industrial Analysis
Gallery™ System Reagents for Environmental and Industrial Analysis
Actual product may vary
Thermo Scientific™
Gallery™ System Reagents for Environmental and Industrial Analysis
Choose from a continuously expanding selection of tests for automated water quality monitoring and nutrient analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
984351TON (nitrate + nitrite) by vanadium reduction system reagents - R2 (500 tests)
984187TON (nitrate + nitrite) by enzymatic reduction system reagents - R1 (275 tests)
984350TON (nitrate + nitrite) by vanadium reduction system reagents - R1 (360 tests)
984357Chromium (VI) system reagents (480 tests)
984358Magnesium system reagents (350 tests)
984362Ammonia system reagents - R1 (2000 tests)
984363Ammonia system reagents - R2 (1300 tests)
984364Chloride system reagents - R1 (500 tests)
984365Chloride system reagents - R1 (2500 tests)
984366Phosphate system reagents - R1 (2300 tests)
984367Phosphate system reagents - R1 (11700 tests)
984368Phosphate system reagents - R2 (3000 tests)
984369TON (nitrate + nitrite) by hydrazine reduction system reagents - R1 (1300 tests)
984370TON (nitrate + nitrite) by hydrazine reduction system reagents - R2 (750 tests)
984371Nitrite system reagents (1000 tests)
984372Nitrite system reagents (5000 tests)
984620Total Hardness system reagents - R1 (1100 tests)
984621Total Hardness system reagents - R2 (975 tests)
984622Total Hardness system reagents - R3 (4300 tests)
984623Alkalinity system reagents - R1 (975 tests)
984624Alkalinity system reagents - R2 (780 tests)
984625Silica system reagents - R1 (970 tests)
984626Silica system reagents - R2 (1950 tests)
984627Silica system reagents - R3 (1950 tests)
984629Fluoride system reagents - R1 (2200 tests)
984630Fluoride system reagents - R2 (4800 tests)
984631Fluoride system reagents - R3 (2200 tests)
984648Sulphate system reagents (1100 tests)
984649Sulphate system reagents (5100 tests)
984652TON (nitrate + nitrite) by hydrazine reduction system reagents - R2 (3400 tests) - for Aquakem only
30 Options
Catalog number 984351
Price (USD)/ Each
-
Request A Quote
Description:
TON (nitrate + nitrite) by vanadium reduction system reagents - R2 (500 tests)
Save time by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for environmental testing, such as water (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use gallery system reagents for environmental analysis, the kits include 1 to 3 reagents in 20mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the discrete analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries. Save time and reduce errors by simplifying your analysis with the Gallery™ system reagents for environmental analysis.
Highlights:
  • All test applications are based on recommended standard methods in water and wastewater analysis of different countries
  • Kit structure includes 1 to 3 reagents in 20mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, are automatically read
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
  • Types of matrices analyzed include clean water, wastewater, soil and sludge digests, effluents, saline and process water
Specifications
TypeTON-V Vanadium Reduction System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionTON (nitrate + nitrite) by vanadium reduction system reagents - R2 (500 tests)
DetectsTON-V R2
No. of Tests500
Test Range0.4 to 50
Quantity4 x 10 mL
Interference Filter540
Unit SizeEach