Gallery™ System Reagents for Environmental and Industrial Analysis
Gallery™ System Reagents for Environmental and Industrial Analysis
Actual product may vary
Gallery™ System Reagents for Environmental and Industrial Analysis
Gallery™ System Reagents for Environmental and Industrial Analysis
Thermo Scientific™
Gallery™ System Reagents for Environmental and Industrial Analysis
Choose from a continuously expanding selection of tests for automated water quality monitoring and nutrient analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeNo. of Tests
984623Alkalinity System Reagent975
984187TON Enzymatic Reduction System Reagent275
984350TON-V Vanadium Reduction System Reagent360
984351TON-V Vanadium Reduction System Reagent500
984357Chromium System Reagent480
984358Magnesium System Reagent350
984362Ammonia System Reagent2000
984363Ammonia System Reagent1300
984364Chloride System Reagent500
984365Chloride System Reagent2500
984366Phosphate System Reagent2300
984367Phosphate System Reagent11700
984368Phosphate System Reagent3000
984369TON System Reagent1300
984370TON System Reagent750
984371TON System Reagent1000
984372TON System Reagent5000
984620Total Hardness System Reagent1100
984621Total Hardness System Reagent975
984622Total Hardness System Reagent4300
984624Alkalinity System Reagent780
984625Silica System Reagent970
984626Silica System Reagent R21950
984627Silica System Reagent R31950
984629Fluoride System Reagent R12200
984630Fluoride System Reagent4800
984631Fluoride System Reagent R32200
984648Sulphate System Reagent1100
984649Sulphate System Reagent5100
984652TON System Reagent3400
Catalog number 984623
Price (USD)
-
Request A Quote
Type:
Alkalinity System Reagent
No. of Tests:
975
Ask our AI about this Product
Save time by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for environmental testing, such as water (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use gallery system reagents for environmental analysis, the kits include 1 to 3 reagents in 20mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the discrete analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries. Save time and reduce errors by simplifying your analysis with the Gallery™ system reagents for environmental analysis.
Highlights:
  • All test applications are based on recommended standard methods in water and wastewater analysis of different countries
  • Kit structure includes 1 to 3 reagents in 20mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, are automatically read
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
  • Types of matrices analyzed include clean water, wastewater, soil and sludge digests, effluents, saline and process water
Specifications
DescriptionAlkalinity system reagents - R1 (975 tests)
DetectsAlkalinity R1
Interference Filter600/ 880
No. of Tests975
Quantity4 x 20 mL
Test Range0 to 400
Sample TypeWater Sample, Soil Sample
TypeAlkalinity System Reagent
Unit SizePack of,4
Explore our full discrete analyzer product offering
Explore our full discrete analyzer product offering
Find additional products for automated wet chemical analysis, including systems for beer, water and enzyme analysis as well as reagents for food and beverage or environmental and industrial analyses.
Learn more ›
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link