1x1 image pixel for data collection
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Actual product may vary
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Thermo Scientific™

Gallery™ Discrete Analyzer

Bring excellent analytical performance to colorimetric, enzymatic and electrochemical measurements with the Thermo Scientific™ Gallery™ Automated Photometric Analyzer.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
98611001Gallery Discrete Analyzer with ECM Unit
98610001Gallery Discrete Analyzer without ECM Unit
Catalog number 98611001
Price (USD)
-
Request A Quote
Description:
Gallery Discrete Analyzer with ECM Unit
Ask our AI about this Product

Designed to increase efficiency for photometric (colorimetric and enzymatic) and electrochemical (pH and conductivity) analysis, the Thermo Scientific™ Gallery™ Discrete Analyzer provides fast, reproducible results in a compact, benchtop design. Its discrete cell technology allows laboratories to measure multiple analytes simultaneously while reducing total analysis and operator time. All necessary analysis steps are automated and low detection levels can be achieved. The analyzer includes a combined sample and reagent disk for a maximum capacity of 90 samples and 30 reagents, with the ability to perform as many as 200 tests/hour. Achieve increased analytical performance in a wide range of applications including food and beverage analysis, environmental testing, and industrial quality control.

Automated operation and reliable technology

  • Continuous access to samples, reagents, and cuvettes without interruption of routine workflows.
  • Integrated platform for photometric and electrochemical analysis can be operated simultaneously.
  • All necessary analysis steps are automated, enabling up to 2 hours walk-away time for the operator.
  • Increased throughput with a sample capacity of up to 200 tests/hour.
  • Dilutions and repeat analyses are handled automatically.
  • Minimal sample pretreatment required.

Robust capability 

  • Large concentration range measurement, with excellent reproducibility.
  • User-specific applications can be developed to meet individual testing needs.
  • Combined sample and reagent disk with up to 90 sample positions on-board.
  • Several blanking possibilities to eliminate sample matrix effects.
  • Ability to achieve detection of very low levels (ppb).
  • Wavelength range from 340nm to 880nm.
  • Easy dilution management.

Compact design and economical

  • System reagents eliminate time-consuming reagent preparation, freeing operators for other tasks.
  • Unique low volume cuvette design for less reagent waste and potentially lower operating costs.
  • Open system – modify existing method or create your own test.
  • Contamination-free analysis using disposable cuvettes.
  • Optional ECM-unit for conductivity and pH.
  • No need for an external source of water.

Versatile and user-friendly 

  • Accommodates a variety of sample cups and primary tubes.
  • No risk of carry-over or requirement to wash glassware.
  • No additional priming or method changeover time.
  • Identification is automated by a barcode reader.
  • Save space: Self-contained benchtop for small footprint.
  • Automatic startup and shutdown protocols.
  • Intuitive graphic user interface.
Specifications
Analytical MethodColorimetric, Enzymatic and Electrochemical With Automated Photometric Analyzer
CalibrationFactor, Bias, Linear, Logit-Log, Spline, Polynomial and Point-to-Point
CapacityMix of 9 to or 18 to Position Sample Racks
CuvettesDiscrete Disposable Cuvettes. On Board Capacity 360 Measurement Cells.
Depth (English)27.5 in.
Depth (Metric)70 cm
DescriptionGallery Discrete Analyzer with ECM Unit
Distilled Water ConsumptionDeionized Water Consumption 1.5 L/hr.
Frequency50/60 Hz +/-5%
Height (English)24.4/51.1 in. (Cover Closed/Cover Open)
Height (Metric)62/130 cm (Cover Closed/Cover Open)
Measurement MethodPhotometric and Electrochemical (ECM)
Operating SystemAutomated Photometric (Colorimetric and Enzymatic) Analysis with The Optional Electrochemical (ECM) Measurement of pH and Electrical Conductivity
Quality Control DetailsReal-Time QC Program with Multiple and Variable Rules. Control Frequency User-Definable. Out-of-Specification Control Results Flagged.
Reagents6-Position Reagent Racks, Max 6 Racks on-Board
Sample DilutionAutomatic Sample Pre-Dilution. Automatic Sample Dilution of Over-Range Tests with Automatic re-Run. Addition of Manual Pre-Dilution Value for Result Calculation.
Throughput200 Tests/hr.
Voltage100/240 V +/-10%
Weight (Metric)85 kg
Width (English)29.5 in.
Width (Metric)75 cm
TypeDiscrete Analyzer
Unit SizeEach
Explore our full discrete analyzer product offering

Explore our full discrete analyzer product offering

Find additional products for automated wet chemical analysis, including systems for beer, water and enzyme analysis as well as reagents for food and beverage or environmental and industrial analyses.

Learn more ›

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Videos

Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
 
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Documents & Downloads

      Certificates

      0 results displayed, search above for a specific certificate

      Scientific Resources

      Product Information

      Share catalog number, name or link