Search Thermo Fisher Scientific
Catalog Number | Electrochemistry Unit (ECM) | Measurement Method | Throughput |
---|---|---|---|
98611004 | Included | Photometric: 340nm to 880nm pH Range: 2-12 pH Conductivity Range: 20 μS/cm – 112 mS/cm | 200 Photometric Tests/hr.; up to 67 pH and conductivity measurements/hr. |
98610004 | Optional | Photometric: 340nm to 880nm | 200 Photometric Tests/hr. |
Confidence in your Results
• Precise temperature control
• Temperature overheating limits
• Accurate incubation time to the seconds
• Superior temperature control and lack of edge effects
Highthroughput Enzyme Assay
• Walkaway solution
• Easy to use, train, and maintain
• Many enzymes, many substrates, many methods—all on a single instrument
• Fully-automated liquid handling (sample, buffer and substrate addition, and mixing)
• Disposable cuvettes (no carryover)
Fully automated workflow
• Easy, automated method development
• Easy method transfer
• Enzyme-specific method parameters all in one central place
Traceability
• Time stamped raw data
• Audit trail
• User administration
Parallel pH Measurement
• ECM Module for parallel pH measurement
• Facilitates method development
• Up to 67 samples pH measurement per hour