Gallery Plus Enzyme Master
Gallery Plus Enzyme Master
Actual product may vary
Thermo Scientific™

Gallery Plus Enzyme Master

Combining robust hardware and new custom-designed software, Gallery Plus Enzyme Master, the best-in-class solutions streamline enzyme assay/activity method development and reliable routine testing—for walkaway efficiency day in and day out.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrochemistry Unit (ECM)Measurement MethodThroughput
98620004OptionalPhotometric: 340nm to 880nm350 Photometric Tests/hr.
98621004IncludedPhotometric: 340nm to 880nm
pH Range: 2-12 pH
Conductivity Range: 20 μS/cm–112 mS/cm
340 Photometric Tests/hr.; up to 67 pH and conductivity measurements/hr.
2 Options
Catalog number 98620004
Price (USD)/ Each
-
Request A Quote
Electrochemistry Unit (ECM):
Optional
Measurement Method:
Photometric: 340nm to 880nm
Throughput:
350 Photometric Tests/hr.

Experience the Thermo Scientific™ Gallery™ Plus Enzyme Master - the first automated discrete analyzers designed specifically for enzyme assay applications. The custom-designed software, reagent additions, incubations, and measurements are fully automated, which saves time, eliminates manual error, and improves method robustness. Reliable instrumentation allows you to facilitate simple method transfers from research and development (R&D) to the quality assurance/quality control floor. With the flexibility and automation of the Thermo Scientific™ Gallery Plus Enzyme Master, you're able to meet your current and future lab needs. The optional Electrochemistry unit (ECM) allows the parallel determination of pH optimal up to 67 samples per hour along with other photometric tests.

Confidence in your Results

  • Precise temperature control
  • Temperature overheating limits
  • Accurate incubation time to the seconds
  • Superior temperature control and lack of edge effects
High throughput Enzyme Assay
  • Walkaway solution, up to 350 photometric tests/hr
  • Easy to use, train, and maintain
  • Many enzymes, many substrates, many methods—all on a single instrument
  • Fully-automated liquid handling (sample, buffer and substrate addition, and mixing)
  • Disposable cuvettes (no carryover)
Fully automated workflow
  • Easy, automated method development
  • Easy method transfer
  • Enzyme-specific method parameters all in one central place
     

Traceability

  • Time stamped raw data
  • Audit trail
  • User administration


Parallel pH Measurement

  • ECM Module for parallel pH measurement
  • Facilitates method development
  • Up to 67 samples pH measurement per hour
Specifications
Sample Size108 Samples
Throughput350 Photometric Tests/hr.
Incubation Temperature25°C to 60°C
Analytical MethodDiscrete Analysis
Reagent CapacityUp to 48
Electrochemistry Unit (ECM)Optional
Measurement MethodPhotometric: 340nm to 880nm
Unit SizeEach