Search by lot number or partial lot number
Type | Photometric Analyzer |
Sample Type | Food and Beverage Sample, Environmental Sample |
Description | Gallery Plus Discrete Analyzer with ECM Unit |
Sample Dilution | Automatic Sample Pre-Dilution. Automatic Sample Dilution of Over-Range Tests with Automatic re-Run. Addition of Manual Pre-Dilution Value for Result Calculation. |
Throughput | Up to 350 Tests/hr. |
Calibration | Factor, Bias, Linear, Logit-Log, Spline, Polynomial and Point-to-Point |
Cuvettes | Discrete Disposable Cuvettes. Capacity 360 Measurement Cells. |
Distilled Water Consumption | Deionized Water Consumption 2.5 L/hr. |
Reagents | 9- or 18-Position Sample Racks, Max 108 Positions (= 6 Racks) on-Board; Reagent Capacity 42 Positions on-Board |
Depth (English) | 27.5 in. |
Depth (Metric) | 70 cm |
Height (English) | 24.4/51.1 in. (Cover Closed/Cover Open) |
Height (Metric) | 62/130 cm (Cover Closed/Cover Open) |
Width (English) | 37 in. |
Width (Metric) | 94 cm |
Frequency | 50/60 Hz +/-5% |
Voltage | 100/240 V +/-10% |
Operating System | Automated Photometric (Colorimetric and Enzymatic) Analysis with The Optional Electrochemical (ECM) Measurement of pH and Electrical Conductivity |
Quality Control Details | Real-Time QC Program with Multiple and Variable Rules. Control Frequency User-Definable. Out-of-Specification Control Results Flagged. |
Measurement Method | Photometric and Electrochemical (ECM) |
Weight (Metric) | 110 kg |
Unit Size | Each |
Type | Photometric Analyzer |
Sample Type | Food and Beverage Sample, Environmental Sample |
Description | Gallery Plus Discrete Analyzer without ECM Unit |
Sample Dilution | Automatic Sample Pre-Dilution. Automatic Sample Dilution of Over-Range Tests with Automatic re-Run. Addition of Manual Pre-Dilution Value for Result Calculation. |
Throughput | Up to 350 Tests/hr. |
Calibration | Factor, Bias, Linear, Logit-Log, Spline, Polynomial and Point-to-Point |
Cuvettes | Discrete Disposable Cuvettes. Capacity 360 Measurement Cells. |
Distilled Water Consumption | Deionized Water Consumption 2.5 L/hr. |
Reagents | 9- or 18-Position Sample Racks, Max 108 Positions (= 6 Racks) on-Board; Reagent Capacity 42 Positions on-Board |
Depth (English) | 27.5 in. |
Depth (Metric) | 70 cm |
Height (English) | 24.4/51.1 in. (Cover Closed/Cover Open) |
Height (Metric) | 62/130 cm (Cover Closed/Cover Open) |
Width (English) | 37 in. |
Width (Metric) | 94 cm |
Frequency | 50/60 Hz +/-5% |
Voltage | 100/240 V +/-10% |
Operating System | Automated Photometric (Colorimetric and Enzymatic) Analysis |
Quality Control Details | Real-Time QC Program with Multiple and Variable Rules. Control Frequency User-Definable. Out-of-Specification Control Results Flagged. |
Measurement Method | Photometric |
Weight (Metric) | 110 kg |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Operating System | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
98621001 | Each | Gallery Plus Discrete Analyzer with ECM Unit | Automated Photometric (Colorimetric and Enzymatic) Analysis with The Optional Electrochemical (ECM) Measurement of pH and Electrical Conductivity | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
98620001 | Each | Gallery Plus Discrete Analyzer without ECM Unit | Automated Photometric (Colorimetric and Enzymatic) Analysis | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|