Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Type | Cuvette |
Description | DECACELL™ Cuvettes |
Color | Clear |
Disposable | Yes |
Capacity (Metric) | 2 to 240 μL |
Quantity | One box of 540 cuvettes with 10 cells each |
For Use With (Equipment) | Thermo Scientific™ Gallery™ and Gallery™ Plus Discrete Analyzers |
For Use With (Application) | Automated Wet Chemical Analysis |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
986540TS | Each | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|