Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product Type | Tubing Cutter |
Material | Plastic |
Diameter (English) Outer | 1/16, 1/8 in. |
Diameter (Metric) Outer | 1.59, 3.175 mm |
For Use With | Upchurch™ standard polymer tubing cutter |
Color | Black |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Color | Material | Product Type | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A-327 | Each | Black | Plastic | Tubing Cutter | 53.89 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|