Ethyl salicylate, 99%, Thermo Scientific Chemicals
Ethyl salicylate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl salicylate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A10101.22
also known as A10101-22
Price (USD)/ Each
27.65
Online exclusive
30.10 
Save 2.45 (8%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
27.65
Online exclusive
30.10 
Save 2.45 (8%)
Add to cart
Ethyl salicylate, 99%, Thermo Scientific Chemicals
Catalog numberA10101.22
Price (USD)/ Each
27.65
Online exclusive
30.10 
Save 2.45 (8%)
-
Add to cart
Chemical Identifiers
CAS118-61-6
IUPAC Nameethyl 2-hydroxybenzoate
Molecular FormulaC9H10O3
InChI KeyGYCKQBWUSACYIF-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.5200-1.5250 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only