Used as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.
Solubility
Soluble in Methanol. Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- V.K. Rastogi; Vaibhav Jain; M. Alcolea Palafox; D.N. Singh; R.A. Yadav. Laser Raman and IR spectra and force fields for 2,4-dichlorobenzonitrile. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2001, 57 (2), 209-216.
- Ibrahim Haddad; Shaun Hurley; C. S. Marvel. Poly(arylene sulfides) with pendant cyano groups as high-temperature laminating resins. Journal of Polymer Science Part A: Polymer Chemistry. 1973, 11 (11), 2793-2811.