4-Amino-2-chlorobenzoic acid, 98%, Thermo Scientific Chemicals
4-Amino-2-chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-2-chlorobenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A10116.14
also known as A10116-14
Price (USD)/ Each
265.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
265.00
Add to cart
4-Amino-2-chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Catalog numberA10116.14
Price (USD)/ Each
265.00
-
Add to cart
Chemical Identifiers
CAS2457-76-3
IUPAC Name4-amino-2-chlorobenzoic acid
Molecular FormulaC7H6ClNO2
InChI KeyMBDUKNCPOPMRJQ-UHFFFAOYSA-N
SMILESNC1=CC=C(C(O)=O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Pale cream to cream to pale brown
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only