Thermo Scientific Chemicals

4-(Bromomethyl)benzenesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: A10120.06
5 g, Each
Thermo Scientific Chemicals

4-(Bromomethyl)benzenesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: A10120.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10120.06
also known as A10120-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66176-39-4
IUPAC Name
4-(bromomethyl)benzene-1-sulfonyl chloride
Molecular Formula
C7H6BrClO2S
InChI Key
QXTQWYZHHMQSQH-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1=CC=C(CBr)C=C1
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥94.0 to ≤106.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
67.0-80.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text