Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Remove inorganic solids, organics, bacteria and pyrogens from your water through the distillation process using Thermo Scientific™ Barnstead™ Classic Stills. Choose from bench or floor mounted models. Available in five capacities to meet your production needs, stills are constructed of copper and bronze with a pure tin coating. The inert nature of tin prevents leaching of contaminants into water. Portable stills allow for quick set-up, while floor model electric stills improve safety with over-temperature protection. Steam stills feature optional automatic controls for unattended operation.
Amperage | 17 A |
Dimensions (L x W x H) | 11 x 23 x 68 in. (28 x 58 x 173 cm) |
Electrical Requirements | 208 V 50/60 Hz |
Frequency | 50/60 Hz |
Phase | 3 |
Voltage | 208 V |
Flow Rate | 2 gal./hr. |
Type | Electric |
Output Flow Rate | 2 gal./hr. |
Model | 2 gal./hr. (204 V) Floor Electric with Stand |
Height (English) | 68 in. |
Height (Metric) | 173 cm |
Width (English) | 23 in. |
Width (Metric) | 58 cm |
Cooling Flow Rate (English) | 16 gal./hr. |
Cooling Flow Rate (Metric) | 61 L/hr. |
Length (English) | 11 in. |
Length (Metric) | 28 cm |
Certifications/Compliance | UL, cUL |
Unit Size | Each |
Amperage | 12 A |
Dimensions (L x W x H) | 10 x 21 x 19 in. (26 x 52 x 49 cm) |
Electrical Requirements | 120 V 50/60 Hz |
Frequency | 50/60 Hz |
Phase | 1 |
Voltage | 120 V |
Flow Rate | 0.5 gal./hr. |
Type | Electric (Portable) |
Output Flow Rate | 0.5 gal./hr. |
Model | 0.5 gal./hr. Portable |
Height (English) | 19 in. |
Height (Metric) | 49 cm |
Width (English) | 21 in. |
Width (Metric) | 52 cm |
Cooling Flow Rate (English) | 4 gal./hr. |
Cooling Flow Rate (Metric) | 15 L/hr. |
Length (English) | 10 in. |
Length (Metric) | 26 cm |
Certifications/Compliance | UL, cUL |
Unit Size | Each |
Amperage | 26 A |
Dimensions (L x W x H) | 11 x 23 x 68 in. (28 x 58 x 173 cm) |
Electrical Requirements | 240 V 50/60 Hz |
Frequency | 50/60 Hz |
Phase | 1 |
Voltage | 240 V |
Flow Rate | 2 gal./hr. |
Type | Electric |
Output Flow Rate | 2 gal./hr. |
Model | 2 gal./hr. (240 V) Floor Electric with Stand |
Height (English) | 68 in. |
Height (Metric) | 173 cm |
Width (English) | 23 in. |
Width (Metric) | 58 cm |
Cooling Flow Rate (English) | 16 gal./hr. |
Cooling Flow Rate (Metric) | 61 L/hr. |
Length (English) | 11 in. |
Length (Metric) | 28 cm |
Certifications/Compliance | UL, cUL |
Unit Size | Each |
Amperage | 57 A |
Dimensions (L x W x H) | 14 x 35 x 77 in. (36 x 89 x 196 cm) |
Electrical Requirements | 240 V 50/60 Hz |
Frequency | 50/60 Hz |
Phase | 1 |
Voltage | 240 V |
Flow Rate | 5 gal./hr. |
Type | Electric |
Output Flow Rate | 5 gal./hr. |
Model | 5 gal./hr. (240 V) Floor Electric with Stand |
Height (English) | 77 in. |
Height (Metric) | 196 cm |
Width (English) | 35 in. |
Width (Metric) | 89 cm |
Cooling Flow Rate (Metric) | 151 L/hr. |
Length (English) | 14 in. |
Length (Metric) | 36 cm |
Certifications/Compliance | UL, cUL |
Unit Size | Each |
Amperage | 36 A |
Dimensions (L x W x H) | 14 x 35 x 77 in. (36 x 89 x 196 cm) |
Electrical Requirements | 208 V 50/60 Hz |
Frequency | 50/60 Hz |
Phase | 3 |
Voltage | 208 V |
Flow Rate | 5 gal./hr. |
Type | Electric |
Output Flow Rate | 5 gal./hr. |
Model | 5 gal./hr. (208 V) Floor Electric with Stand |
Height (English) | 77 in. |
Height (Metric) | 196 cm |
Width (English) | 35 in. |
Width (Metric) | 89 cm |
Cooling Flow Rate (English) | 40 gal./hr. |
Cooling Flow Rate (Metric) | 151 L/hr. |
Length (English) | 14 in. |
Length (Metric) | 36 cm |
Certifications/Compliance | UL, cUL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Type | Output Flow Rate | Electrical Requirements | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A1013-C | Each | Electric | 2 gal./hr. | 208 V 50/60 Hz | 13,800.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A1007 | Each | Electric (Portable) | 0.5 gal./hr. | 120 V 50/60 Hz | 4,716.00 Online price 5,240.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A1013-B-61 | Each | Electric | 2 gal./hr. | 240 V 50/60 Hz | 13,600.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A1015-B-61 | Each | Electric | 5 gal./hr. | 240 V 50/60 Hz | 21,500.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A1015-C | Each | Electric | 5 gal./hr. | 208 V 50/60 Hz | 21,500.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Portable Stills:
Floor Model Electric Still:
Steam Still:
Ordering Information:
Automated control systems are available for all stills: G2100 (for 1 gal./hr., 2 gal./hr. models), G2110 (5 gal./hr.), G2125 (10 gal./hr.) and G2010 (10 gal./hr.). Order separately.
This product is not available in all regions.
Includes:
Floor stand (exept for Portable Model A1007).
Requires:
Storage reservoir, UV light assembly and Ventgard Air Filter (order separately)
Recommended for:
Buffer/reagent prep, glassware washing, dishwasher feed