Thermo Scientific Chemicals

DL-Phenylalanine, 99%

Catalog number: A10132.36
500 g, Each
Thermo Scientific Chemicals

DL-Phenylalanine, 99%

Catalog number: A10132.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10132.36
also known as A10132-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
150-30-1
IUPAC Name
2-amino-3-phenylpropanoic acid
Molecular Formula
C9H11NO2
InChI Key
COLNVLDHVKWLRT-UHFFFAOYNA-N
SMILES
NC(CC1=CC=CC=C1)C(O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.

Solubility
Soluble in water. Insoluble in ethyl ether, benzene.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text